Skip to Content
Merck
All Photos(1)

Documents

SML0577

Sigma-Aldrich

Cucurbitacin E

≥95% (HPLC)

Synonym(s):

α-Elaterin, α-Elaterine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H44O8
CAS Number:
Molecular Weight:
556.69
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D -60 to -75°, c = 0.7 (CDCl3)

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

InChI

1S/C32H44O8/c1-17(33)40-27(2,3)13-12-23(36)32(9,39)25-21(35)15-29(6)22-11-10-18-19(14-20(34)26(38)28(18,4)5)31(22,8)24(37)16-30(25,29)7/h10,12-14,19,21-22,25,34-35,39H,11,15-16H2,1-9H3/b13-12+/t19-,21-,22+,25+,29+,30-,31+,32+/m1/s1

InChI key

NDYMQXYDSVBNLL-MUYMLXPFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cucurbitacin E has been used as a cofilin inhibitor. It is also used as a F-actin stabilizer to prevent membrane-associated periodic skeleton (MPS) loss and protect from axonal fragmentation.

Biochem/physiol Actions

Cucurbitacin E is a potent inhibitor of actin depolymerization. Cucurbitacin E is more active than jasplakinolide, and has a different mechanism of action, binding to a different site. Cucurbitacin E binds specifically to filamentous actin (F-actin) forming a covalent bond at residue Cys257, but not to monomeric actin (G-actin), stabilizing F-actin, without affecting actin polymerization or nucleation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three-dimensional structural analysis reveals a Cdk5-mediated kinase cascade regulating hepatic biliary network branching in zebrafish
Dimri M, et al.
Development, 144(14), 2595-2605 (2017)
Lamice Habib et al.
International journal of pharmaceutics, 448(1), 313-319 (2013-03-26)
Cucurbitacin E (Cuc E), highly oxygenated triterpene from cucurbitaceae family, was demonstrated to possess anti-proliferative, anti-inflammatory and anti-oxidant activities. Here, we studied the effect of Cuc E on the properties of the phospholipid membrane. Large unilamellar vesicles, with and without
Chao-Ming Hung et al.
International journal of molecular sciences, 14(8), 17147-17156 (2013-08-24)
Human oral squamous cell carcinoma (OSCC) is a common form of malignant cancer, for which radiotherapy or chemotherapy are the main treatment methods. Cucurbitacin E (CuE) is a natural compound previously shown to be an antifeedant as well as a
Xiaolong He et al.
Oncology letters, 13(6), 4905-4910 (2017-06-11)
Cucurbitacin E is an important member of the cucurbitacin family and exhibits inhibitory effects in various types of cancer. Cucurbitacin is a potential antineoplastic drug; however, its anticancer effect in human prostate cancer (PC) remains unknown. The aim of the
Tao Zhang et al.
Breast cancer research and treatment, 135(2), 445-458 (2012-07-31)
Tumor metastasis is the main cause of cancer-related deaths of patients. Breast cancer is highly malignant with considerable metastatic potential, which urges the necessity for developing novel potential drug candidate to prevent tumor metastasis. Here, we report our finding with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service