Skip to Content
Merck
All Photos(1)

Documents

SML0505

Sigma-Aldrich

Tienilic Acid

≥98% (HPLC)

Synonym(s):

Ticrynafen, Tienylic acid, [2,3-Dichloro-4-(2-thienylcarbonyl)phenoxy]-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8Cl2O4S
CAS Number:
Molecular Weight:
331.17
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)COc1ccc(c(Cl)c1Cl)C(=O)c2cccs2

InChI

1S/C13H8Cl2O4S/c14-11-7(13(18)9-2-1-5-20-9)3-4-8(12(11)15)19-6-10(16)17/h1-5H,6H2,(H,16,17)

InChI key

AGHANLSBXUWXTB-UHFFFAOYSA-N

Biochem/physiol Actions

Tienilic Acid (Ticrynafen) is a P450 inhibitor, a specific suicide substrate for CYP2C9 and CYP2C10. It was once used as a loop diuretic drug with uric acid-lowering (uricosuric) actvity, but was removed from market because of hepatotoxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahendra Ramesh Shiradkar et al.
Bioorganic & medicinal chemistry, 15(19), 6397-6406 (2007-07-24)
Based on the earlier results of our in-house database and compound library, a series of novel clubbed thienyl triazoles was designed which may emerge as potential cdk5/p25 inhibitors, for the treatment of Alzheimer's disease. A benign synthesis was planned so
Takayoshi Nishiya et al.
Toxicology letters, 183(1-3), 81-89 (2008-11-11)
Tienilic acid is reported to be converted into electrophilic metabolites by cytochrome P450 (CYP) in vitro. In vivo, however, the metabolites have not been detected and their effect on liver function is unknown. We previously demonstrated that tienilic acid decreased
M Pilar López-García et al.
Biochemical pharmacology, 70(12), 1870-1882 (2005-11-01)
Drug-induced autoimmune hepatitis is among the most severe hepatic idiosyncratic adverse drug reactions. Considered multifactorial, the disease combines immunological and metabolic aspects, the latter being to date much better known. As for many other model drugs, studies on tienilic acid
Hideo Takakusa et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(5), 816-823 (2008-01-30)
The metabolic activation of a drug to an electrophilic reactive metabolite and its covalent binding to cellular macromolecules is considered to be involved in the occurrence of idiosyncratic drug toxicity (IDT). As a cellular defense system against oxidative and electrophilic
S Poli-Scaife et al.
Biochemistry, 36(42), 12672-12682 (1997-10-23)
Purified recombinant human liver cytochrome P450 2C9 was produced, from expression of the corresponding cDNA in yeast, in quantities large enough for UV-visible and 1H NMR experiments. Its interaction with several substrates (tienilic acid and two derivatives, lauric acid and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service