Skip to Content
Merck
All Photos(1)

Key Documents

SML0435

Sigma-Aldrich

AF-DX 116

≥98% (HPLC)

Synonym(s):

11-[[2-[(Diethylamino)methyl]-1-piperidinyl]acetyl]-5,1 1-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one, Otenzepad

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31N5O2
CAS Number:
Molecular Weight:
421.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution; warmed)

storage temp.

room temp

SMILES string

CCN(CC)CC1CCCCN1CC(=O)N2c3ccccc3C(=O)Nc4cccnc24

InChI

1S/C24H31N5O2/c1-3-27(4-2)16-18-10-7-8-15-28(18)17-22(30)29-21-13-6-5-11-19(21)24(31)26-20-12-9-14-25-23(20)29/h5-6,9,11-14,18H,3-4,7-8,10,15-17H2,1-2H3,(H,26,31)

InChI key

UBRKDAVQCKZSPO-UHFFFAOYSA-N

Application

AF-DX 116 has been used as an M2 muscarinic acetylcholine receptor (M2AChR) selective antagonist to study the influence of M2AChR in the regulation of both basal and carbachol-stimulated long-term potentiation at the dentate gyrus (DG-LTP) and pro-nerve growth factor (proNGF) secretion in hippocampal cells of young diabetic rats.

Biochem/physiol Actions

AF-DX 116 is a selective M2 muscarinic acetylcholine receptor antagonist.
AF-DX 116 raises blood pressure and heart rate in a rat model of hypotension induced by repeated cold stress. Thus, it may be used as a therapeutic for hypotension related to vagotonia type autonomic dysfunction.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leticia Ramírez-Lugo et al.
Neurobiology of learning and memory, 79(2), 184-193 (2003-02-20)
A number of studies have implicated cholinergic activity in the mediation of learning and memory processes. However, the specific role of muscarinic receptors in memory formation mechanisms is less known. The aim of the present study is to evaluate the
Alfredo González et al.
BMC neuroscience, 5, 23-23 (2004-07-15)
In fish, melanin pigment granules in the retinal pigment epithelium disperse into apical projections as part of the suite of responses the eye makes to bright light conditions. This pigment granule dispersion serves to reduce photobleaching and occurs in response
Makoto Ukai et al.
European journal of pharmacology, 492(2-3), 183-187 (2004-06-05)
The effects of anticholinergic drugs selective for muscarinic receptor subtypes on prepulse inhibition of acoustic startle response were determined in mice. The prepulse inhibition is associated with sensorimotor information processing in the brain. The anticholinergic agent scopolamine (0.3 mg/kg, s.c.)
Julia B R Pfeiffer et al.
American journal of veterinary research, 68(3), 313-322 (2007-03-03)
To describe the in vitro effects of bethanechol on contractility of smooth muscle preparations from the small intestines of healthy cows and define the muscarinic receptor subtypes involved in mediating contraction. Tissue samples from the duodenum and jejunum collected immediately
Máriton D Santos et al.
The Journal of pharmacology and experimental therapeutics, 304(1), 254-265 (2002-12-20)
This study was designed to investigate the effects of the cholinesterase inhibitors soman and pyridostigmine bromide (PB) on synaptic transmission in the CA1 field of rat hippocampal slices. Soman (1-100 nM, 10-15 min) decreased the amplitude of GABAergic postsynaptic currents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service