Skip to Content
Merck
All Photos(3)

Key Documents

O100

Sigma-Aldrich

Oxotremorine M

solid

Synonym(s):

N,N,N,-Trimethyl-4-(2-oxo-1-pyrrolidinyl)-2-butyn-1-ammonium iodide, Oxotremorine methiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19IN2O
CAS Number:
Molecular Weight:
322.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: 24 mg/mL
ethanol: 4.6 mg/mL

storage temp.

2-8°C

SMILES string

[I-].C[N+](C)(C)CC#CCN1CCCC1=O

InChI

1S/C11H19N2O.HI/c1-13(2,3)10-5-4-8-12-9-6-7-11(12)14;/h6-10H2,1-3H3;1H/q+1;/p-1

InChI key

VVLMSCJCXMBGDI-UHFFFAOYSA-M

Application

Oxotremorine M has been used as a muscarinic acetylcholine receptor (mAchR) agonist:
  • to study its acute effects on the movement of Caenorhabditis elegans
  • to treat hippocampal neurons for studying fibroblast growth factor receptor 1 (FGFR1) phosphorylation
  • to induce bladder overactivity in rats

Biochem/physiol Actions

Oxotremorine M is a nonselective muscarinic acetylcholine receptor agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) and Acetylcholine Receptors (Nicotinic) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Correa et al.
PLoS genetics, 8(11), e1003015-e1003015 (2012-11-21)
Caenorhabditis elegans male copulation requires coordinated temporal-spatial execution of different motor outputs. During mating, a cloacal circuit consisting of cholinergic sensory-motor neurons and sex muscles maintains the male's position and executes copulatory spicule thrusts at his mate's vulva. However, distinct
Li Hu et al.
Scientific reports, 10(1), 17581-17581 (2020-10-18)
SPARC-deficient mice have been shown to exhibit impaired glucose tolerance and insulin secretion, but the underlying mechanism remains unknown. Here, we showed that SPARC enhanced the promoting effect of Muscarinic receptor agonist oxotremorine-M on insulin secretion in cultured mouse islets.
Simona Dedoni et al.
Journal of neurochemistry, 122(1), 58-71 (2012-04-27)
Type I interferons (IFNs) have been shown to act on neurons and to cause neuronal damage through mechanisms not completely defined. Here, we investigated the effects of type I IFNs on brain-derived neurotrophic factor (BDNF)-induced TrkB receptor signaling and neurotrophic
M Gillard et al.
Molecular pharmacology, 32(1), 100-108 (1987-07-01)
We compared the binding characteristics of muscarinic receptors labeled by [3H]oxotremorine-M ([3H]oxo-M) in homogenates of brain cortex and heart from rat. In both tissues [3H]oxo-M bound, with the same KD (6.5 nM), to a fraction of the receptors labeled by
Yoshihiro Matsumoto et al.
Urology, 75(4), 862-867 (2010-02-17)
To investigate the effects of various muscarinic acetylcholine receptor (mAChR) antagonists, including selective M2 and M3 mAChR antagonists, on bladder overactivity. It has been proposed that the urothelium modulates the activity of bladder afferent pathways. However, the differential roles of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service