Skip to Content
Merck
All Photos(1)

Documents

SML0430

Sigma-Aldrich

PR-619

≥95% (HPLC)

Synonym(s):

2,6-Diamino-3,5-dithiocyanopyridine, Thiocyanic acid C,C′-(2,6-diamino-3,5-pyridinediyl) ester; 2,6-Diaminopyridine-3,5-bis(thiocyanate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N5S2
CAS Number:
Molecular Weight:
223.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

−20°C

SMILES string

Nc1nc(N)c(SC#N)cc1SC#N

InChI

1S/C7H5N5S2/c8-2-13-4-1-5(14-3-9)7(11)12-6(4)10/h1H,(H4,10,11,12)

InChI key

ZXOBLNBVNROVLC-UHFFFAOYSA-N

Application

PR-619 has been used:
  • as a component of lysis buffer and also as a deubiquitinase inhibitor to treat proteins derived from SILAC-labeled Jurkat cells
  • as a pan-deubiquitinase inhibitor to study its effect on adenoassociated virus (AAV) transduction
  • in radioimmunoprecipitation assay buffer (RIPA) buffer used for ubiquitination assays

Biochem/physiol Actions

PR-619 is a cell permeable broad spectrum deubiquitylating enzymes (DBUs) inhibitor. PR-619 induces the accumulation of polyubiquitylated proteins in cells without directly affecting proteasome activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Wang et al.
Animal reproduction science, 187, 64-73 (2017-10-17)
This study was undertaken to investigate the dynamics of protein ubiquitination in pig gametes and their micro-environments, as well as to explore the action of deubiquitinases (DUBs) in sperm-oocyte binding. Protein ubiquitination states were evaluated by in the ejaculated sperm
Hirotaka Takahashi et al.
Biomedicines, 8(6) (2020-06-10)
Protein ubiquitinations play pivotal roles in many cellular processes, including homeostasis, responses to various stimulations, and progression of diseases. Deubiquitinating enzymes (DUBs) remove ubiquitin molecules from ubiquitinated proteins and cleave the polyubiquitin chain, thus negatively regulating numerous ubiquitin-dependent processes. Dysfunctions
Anne Garreau et al.
Journal of immunology (Baltimore, Md. : 1950), 199(8), 2758-2766 (2017-09-08)
Themis is a new component of the TCR signaling machinery that plays a critical role during T cell development. The positive selection of immature CD4
Cytosolic iron-sulfur assembly is evolutionarily tuned by a cancer-amplified ubiquitin ligase
Weon JL, et sl.
Molecular Cell, 69(1), 113-125 (2018)
Large-scale identification of ubiquitination sites by mass spectrometry
Udeshi ND, et al.
Nature Protocols, 8(10), 1950-1950 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service