Skip to Content
Merck
All Photos(1)

Key Documents

SML0315

Sigma-Aldrich

CORM-A1

≥95% (NMR)

Synonym(s):

Sodium boranocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3BNa2O2
CAS Number:
Molecular Weight:
103.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (NMR)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: >15 mg/mL

storage temp.

room temp

SMILES string

[Na+].[Na+].[BH3-]C([O-])=O

InChI

1S/CH4BO2.2Na/c2-1(3)4;;/h2H3,(H,3,4);;/q-1;2*+1/p-1

InChI key

SOFPSQNQOQPAAJ-UHFFFAOYSA-M

Application

CORM-A1 has been used:
  • to deliver carbon monoxide (CO) and to test its cytoprotection in yeast and primary astrocytes culture during oxidative stress
  • as CO donor in murine macrophages J774A.1 cells to test its effect on cellular β-endorphins elevation
  • to test its effect on mitophagy activation in retinal ganglion cells

Biochem/physiol Actions

CORM-A1 is a water-soluble carbon monoxide (CO) releasing molecule that can be used to study the effects of CO on cellular systems. Carbon monoxide (CO), produced during the degradation of heme by the enzyme heme oxygenase is an important gaseous signaling mediator in mammalian cells CORM-A1 has anti-oxidant and anti-inflammatory activity.
It mediates the release of CO in a pH and temperature-dependent manner, thus favoring mild vasorelaxation and hypotension. During oxidative stress, CORM-A1 is reported to provide cytoprotection in astrocyte primary cultures. This boron-containing CORM promotes autophagy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara R Oliveira et al.
The European journal of neuroscience, 52(1), 2771-2780 (2020-03-14)
Previous studies about the modulation of the vasculature by CO were performed exclusively in male or sexually immature animals. Understanding the sex differences regarding systemic drug processing and pharmacodynamics is an important feature for safety assessment of drug dosing and
Helena Parfenova et al.
American journal of physiology. Heart and circulatory physiology, 315(4), H978-H988 (2018-07-22)
Neonatal asphyxia leads to cerebrovascular disease and neurological complications via a mechanism that may involve oxidative stress. Carbon monoxide (CO) is an antioxidant messenger produced via a heme oxygenase (HO)-catalyzed reaction. Cortical astrocytes are the major cells in the brain
Cláudia Figueiredo-Pereira et al.
FEMS yeast research, 19(5) (2019-07-26)
Autophagy is an autodigestive process, promoting cytoprotection by the elimination of dysfunctional organelles, misfolded proteins and toxic aggregates. Carbon monoxide (CO) is an endogenous gasotransmitter that under low concentrations prevents cell death and inflammation. For the first time, the role
Myrna Constantin et al.
International journal of hypertension, 2012, 859235-859235 (2012-04-21)
Heme oxygenase (HO), a catabolic enzyme, provides the rate-limiting step in the oxidative breakdown of heme, to generate carbon monoxide (CO), iron, and biliverdin-IXα. Induction of the inducible form, HO-1, in tissues is generally regarded as a protective mechanism. Over
Jianxiong Liu et al.
Pediatric research, 82(5), 881-887 (2017-07-01)
BackgroundThe potential contribution of sex-related variables to cerebrovascular functions in neonates remains elusive. Newborn piglets provide a translationally relevant model for studying the effects of seizures in the neonatal brain. The present study investigated whether sex differences contribute to cerebrovascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service