Skip to Content
Merck
All Photos(1)

Documents

SML0247

Sigma-Aldrich

Rizatriptan benzoate salt

≥98% (HPLC)

Synonym(s):

MK-462, N,N-Dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethylamine benzoate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N5 · C7H6O2
CAS Number:
Molecular Weight:
391.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: ≥5 mg/mL

storage temp.

room temp

SMILES string

OC(=O)c1ccccc1.CN(C)CCc2c[nH]c3ccc(Cn4cncn4)cc23

InChI

1S/C15H19N5.C7H6O2/c1-19(2)6-5-13-8-17-15-4-3-12(7-14(13)15)9-20-11-16-10-18-20;8-7(9)6-4-2-1-3-5-6/h3-4,7-8,10-11,17H,5-6,9H2,1-2H3;1-5H,(H,8,9)

InChI key

JPRXYLQNJJVCMZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Rizatriptan is a serotonin 5HT-1B/1D-receptor agonist used to treat migraine.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert MacNeill et al.
Bioanalysis, 4(10), 1163-1173 (2012-06-02)
In the context of matrix effects in bioanalytical LC-MS/MS, a speculative link between free cholesterol, the recoveries of the compound from three common extraction procedures, and response enhancement was qualitatively investigated. Injections on-column of cholesterol both in solution and extracts
Zhao Dong et al.
Brain research, 1368, 151-158 (2010-10-26)
The trigeminovascular nociception induced by electrical stimulation of the dura mater surrounding the superior sagittal sinus in anesthetized animals has been widely used as a model for investigation of the pathophysiology of vascular headache such as migraine. However, little is
Types of colitis based on histology.
Muhammed Sherid et al.
Disease-a-month : DM, 57(9), 457-489 (2011-09-29)
Anthony J Rodgers et al.
Headache, 51(3), 356-368 (2010-11-03)
To examine total migraine freedom (TMF), defined as pain freedom and absence of associated symptoms, using rizatriptan clinical trial data and to explore advantages of TMF as a single primary composite efficacy endpoint. The FDA has set a higher regulatory
Iain P Fraser et al.
Headache, 52(4), 625-635 (2012-02-01)
To evaluate the pharmacokinetic profile and tolerability of single doses of rizatriptan oral disintegrating tablets (ODTs) in pediatric migraineurs. Acute migraine treatment options for children are limited despite a rising migraine prevalence. No triptans are approved in those under 12

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service