Skip to Content
Merck
All Photos(1)

Documents

N1415

Sigma-Aldrich

Naratriptan hydrochloride

≥98% (HPLC)

Synonym(s):

Amerge, GR-85548A, N-Methyl-3-(1-methyl-4-piperidinyl)-1H-indole-5-ethanesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N3O2S ·HCl
CAS Number:
Molecular Weight:
371.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥10 mg/mL

originator

GlaxoSmithKline

storage temp.

room temp

SMILES string

Cl.CNS(=O)(=O)CCc1ccc2[nH]cc(C3CCN(C)CC3)c2c1

InChI

1S/C17H25N3O2S.ClH/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14;/h3-4,11-12,14,18-19H,5-10H2,1-2H3;1H

InChI key

AWEZYKMQFAUBTD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Naratriptan hydrochloride is a triptan drug that is used to relieve acute migraine headaches. It has poor bioavailability, short half-life, and the incidence of migraine recurrence therefore, it is occasionally used.
Naratriptan hydrochloride is a serotonin 5-HT1B/1D receptor agonist.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Formulation and evaluation of thermoreversible mucoadhesive in-situ gel for intranasal delivery of naratriptan hydrochloride
Shelke S, et al.
Journal of drug delivery science and technology, 238-244 (2015)
Estimation of Naratriptan Hydrochloride in Bulk and Formulation by First Order Derivative UV-Spectrophotometric Methods
Borse JS and Shirkhedkar AA
Journal of Applied Pharmaceutical Science, 227-227 (2012)

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service