Skip to Content
Merck
All Photos(1)

Key Documents

SMB00094

Sigma-Aldrich

Catalposide

≥95% (LC/MS-ELSD)

Synonym(s):

Hydroxybenzoyl catalpol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O12
CAS Number:
Molecular Weight:
482.43
EC Number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC.OC[C@H]1O[C@@H](O[C@@H]2OC=C[C@H]3[C@H](OC(=O)c4ccc(O)cc4)[C@@H]5O[C@]5(CO)[C@@H]23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C22H26O12.C2H6/c23-7-12-14(26)15(27)16(28)21(31-12)33-20-13-11(5-6-30-20)17(18-22(13,8-24)34-18)32-19(29)9-1-3-10(25)4-2-9;1-2/h1-6,11-18,20-21,23-28H,7-8H2;1-2H3/t11-,12-,13-,14-,15+,16-,17+,18+,20+,21+,22-;/m1./s1

InChI key

HDJSRSCFWHTJOZ-BNQCZGKOSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Deane Bowers
Journal of chemical ecology, 29(10), 2359-2367 (2003-12-20)
The growth and survival of the Catalpa sphinx, Ceratomia catalpae (Sphingidae), were measured on five different species of Catalpa: C. bignonioides, C. bungeii, C. fargeseii, C. ovata, and C. speciosa. Larval growth varied significantly among these host plant species; however
Ji Hae Lee et al.
Biochemical and biophysical research communications, 422(4), 568-572 (2012-05-16)
Peroxisome proliferator-activated receptor-alpha (PPARα) is a nuclear receptor that regulates the expression of genes related to cellular lipid uptake and oxidation. Thus, PPARα agonists may be important in the treatment of hypertriglyceridemia and hepatic steatosis. In this study, we demonstrated
Sang-Wook Kim et al.
Inflammatory bowel diseases, 10(5), 564-572 (2004-10-09)
Certain irinoid-producing plants have been used as herbal anti-inflammatory remedies. Here we evaluated whether catalposide (CATP), a single compound isolated from irinoid-producing plant Catalpa ovata, has a potential for preventing or ameliorating diseases characterized by mucosal inflammation. Preliminary microarray-based gene
Mi Kyoung Moon et al.
Toxicology letters, 145(1), 46-54 (2003-09-10)
Catalposide, the major iridoid glycoside isolated from the stem bark of Catalpa ovata G. Don (Bignoniaceae) has been shown to possess anti-microbial, anti-tumoral, and anti-inflammatory properties. Heme oxygenase-1 (HO-1) is a stress response protein and is known to play a
Hyuncheol Oh et al.
Planta medica, 68(8), 685-689 (2002-09-11)
Catalposide (1) and two related iridoids were isolated from the stem of Catalpa ovata (Bignoniaceae) by bioassay guided fractionation. Catalposide (1) significantly inhibited the production of nitric oxide (NO) in lipopolysaccharide (LPS)-stimulated RAW 264.7 macrophages in a dose-dependent manner. RT-PCR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service