Skip to Content
Merck
All Photos(1)

Key Documents

SMB00078

Sigma-Aldrich

Arctiin

≥95% (LC/MS-ELSD)

Synonym(s):

Arctigenin-4-glucoside, Arctigenin 4-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34O11
CAS Number:
Molecular Weight:
534.55
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1ccc(C[C@H]2COC(=O)[C@@H]2Cc3ccc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c(OC)c3)cc1OC

InChI

1S/C27H34O11/c1-33-18-6-4-14(10-20(18)34-2)8-16-13-36-26(32)17(16)9-15-5-7-19(21(11-15)35-3)37-27-25(31)24(30)23(29)22(12-28)38-27/h4-7,10-11,16-17,22-25,27-31H,8-9,12-13H2,1-3H3/t16-,17+,22+,23+,24-,25+,27+/m0/s1

InChI key

XOJVHLIYNSOZOO-SWOBOCGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source, such as Forsythiae Fructus and Arctium lappa L. Arctiin is a lignan.

Biochem/physiol Actions

Arctiin has shown to be a suppressor of cyclin D1 protein expression in multiple types of human tumor cells, such as osteosarcoma, lung, colorectal, cervical, breast, melanoma, and prostate cancer. Arctiin has also shown antiviral activity against influenza A when used alone or in combination with oseltamivir. Arctiin plays an important role in the process of inflammation, by preventing the release of nitric oxide, prostaglandin E2, TNF-α (tumor necrosis factor − α), interleukin (IL)-1β and IL-6. A study using rat model shows that arctiin possess anti-diabetic activity and might serve as an inhibitor of diabetic retinopathy.
Arctiin has shown to be a suppressor of cyclin D1 protein expression in multiple types of human tumor cells, such as osteosarcoma, lung, colorectal, cervical, breast, melanoma, and prostate cancer. Arctiin has also shown antiviral activity against influenza A when used along or in combination with oseltamivir.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan He et al.
Planta medica, 78(8), 800-806 (2012-04-14)
The pharmacokinetic profile of arctiin, the major active lignan in fruits of Arctium lappa L., was investigated. Its main meta"bolite arctigenin was identified by an LC-MS method, and an HPLC-UV technique was developed for the simultaneous quantification of the metabolite
Anti-inflammatory function of arctiin by inhibiting COX-2 expression via NF-κB pathways
Lee S, et al.
Journal of Inflammation, 8(1), 16-16 (2011)
Xudong Yuan et al.
Journal of pharmaceutical and biomedical analysis, 39(3-4), 697-704 (2005-06-11)
A high performance liquid chromatography (HPLC) method was developed for the simultaneous determination of arctiin, chlorogenic acid and glycyrrhizin in the tablets of a Chinese proprietary medicine named, "Yin Qiao Jie Du Pian". The analysis was performed by a reverse
Mu Hong Lee et al.
The Journal of pharmacy and pharmacology, 59(1), 123-130 (2007-01-18)
In this study, we aimed to investigate the anti-inflammatory activity, antinociceptive activity and the action mechanism of Trachelospermi caulis extract. The anti-inflammatory effects were investigated using arachidonic acid, 12-O-tetradecanoylphorbol 13-acetate or carrageenan-induced oedema assays. Antinociceptive activity, using the acetic acid-induced
Heng Chen et al.
European journal of pharmacology, 875, 173053-173053 (2020-03-07)
RIPK1/RIPK3/MLKL (Receptor-interacting protein kinase 1/Receptor-interacting protein kinase 3/Mixed lineage kinase domain-like protein) pathway-mediated necroptosis contributes to myocardial ischemia/reperfusion (I/R) injury, and Arctiin can prevent myocardial fibrosis and hypertrophy. This study aims to explore the effect of Arctiin on myocardial I/R

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service