Skip to Content
Merck
All Photos(1)

Documents

SMB00075

Sigma-Aldrich

(−)-Arctigenin

≥95% (LC/MS-ELSD)

Synonym(s):

2(3H)-Furanone, (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]dihydro-3-[(4-hydroxy-3-methoxyphenyl)methyl]-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O6
CAS Number:
Molecular Weight:
372.41
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc(C[C@@H]2[C@H](COC2=O)Cc3ccc(OC)c(OC)c3)ccc1O

InChI

1S/C21H24O6/c1-24-18-7-5-13(11-20(18)26-3)8-15-12-27-21(23)16(15)9-14-4-6-17(22)19(10-14)25-2/h4-7,10-11,15-16,22H,8-9,12H2,1-3H3/t15-,16+/m0/s1

InChI key

NQWVSMVXKMHKTF-JKSUJKDBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Arctigenin is a lignan found in the root of Arctium lappa L. It has been used in pharmacologic, as well as cosmetic, research and has demonstrated several antiviral and anticancer effects.

Biochem/physiol Actions

Dibenzylbutyrolactone ligand, natural product, viral integrase and topoisomerase II inhibitor, antioxidant, antiviral, anti-inflammatory, immunomodulator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengrong Sun et al.
Planta medica, 77(2), 141-145 (2010-08-19)
The involvement of unfolded protein response (UPR) activation in tumor survival and resistance to chemotherapies suggests a new anticancer strategy targeting UPR pathway. Arctigenin, a natural product, has been recently identified for its antitumor activity with selective toxicity against cancer
Ji Yun Lee et al.
Archives of pharmacal research, 33(6), 947-957 (2010-07-08)
We previously reported that arctigenin, a phenylpropanoid dibenzylbutyrolactone lignan isolated from Forsythia koreana, exhibits anti-inflammatory, antioxidant, and analgesic effects in animal models. In addition, arctigenin inhibited eosinophil peroxidase and activated myeloperoxidase in inflamed tissues. In this study, we tested the
Suresh Awale et al.
Cancer research, 66(3), 1751-1757 (2006-02-03)
Tumor cells generally proliferate rapidly and the demand for essential nutrients as well as oxygen always exceeds the supply due to the unregulated growth and the insufficient and inappropriate vascular supply. However, cancer cells show an inherent ability to tolerate
Vivek Swarup et al.
The Journal of antimicrobial chemotherapy, 61(3), 679-688 (2008-01-31)
OBJECTIVES; To evaluate therapeutic efficacy of arctigenin in an experimental model of Japanese encephalitis (JE). Four- to 5-week-old BALB/c mice of either sex were infected intravenously with lethal dose of 3 x 10(5) pfu of Japanese encephalitis virus (JEV). By
Jong-Sik Jin et al.
Biological & pharmaceutical bulletin, 30(5), 904-911 (2007-05-03)
Plant lignans, such as pinoresinol diglucoside, secoisolariciresinol diglucoside and arctiin, are metabolized to mammalian lignans, enterolactone or enterodiol, by human intestinal bacteria. Their metabolic processes include deglucosylation, ring cleavage, demethylation, dehydroxylation and oxidation. Here we isolated an intestinal bacterium capable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service