Skip to Content
Merck
All Photos(1)

Documents

SAB4700448

Sigma-Aldrich

Monoclonal Anti-MYC-FITC , (C-terminal) antibody produced in mouse

clone 9E10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Cellular myelocytomatosis oncogene, Anti-c-Myc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E10, monoclonal

form

buffered aqueous solution

species reactivity

human, fusion proteins in all species

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

The cellular myelocytomatosis (c-myc) gene mapped to human chromosome 8q24, is the cellular homologue of the v-myc gene originally isolated from an avian myelocytomatosis virus. C-myc is a member of MYC gene family. c-Myc gene codes for basic helix-loop-helix/leucine zipper (bHLH/LZ) transcription factor that regulates the G1-S cell cycle transition. Increased expression of c-myc is observed in variety of human cancerous tissue.

Immunogen

Synthetic peptide sequence (AEEQKLISEEDLL) corresponding to the C-terminal region of human c-Myc

Application

Monoclonal Anti-MYC-FITC , (C-terminal) antibody produced in mouse has been used in immunofluorescence and flow cytometry.

Biochem/physiol Actions

The cellular myelocytomatosis (c-myc) oncogene plays a vital role in cellular proliferation, differentiation, apoptosis and acts as a transcriptional regulator of gene expression. c-Myc expression is essential and sufficient to assist most of the cells to enter DNA synthetic (S) phase of the cell cycle. The encoded protein plays a crucial role in vasculogenesis and angiogenesis during cancer development and progression. c-Myc interacts with its binding partner Max and activates the transcription of growth promoting genes such as cyclin D2, ornithine decarboxylase, and transcription factor E2F1 and it also represses the transcription of multiple genes, especially p21 and p27, by binding to the transcription initiator element (Inr) in a complex with Max and either Sp1 or Miz1 (transcription factors).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoki Maeda et al.
Journal of immunology (Baltimore, Md. : 1950), 201(1), 202-214 (2018-05-18)
The lymphotoxin (LT)/LTβ receptor (LTβR) axis is crucial for the regulation of immune responses and development of lymphoid tissues in mammals. Despite the importance of this pathway, the existence and function of LT and LTβR remain obscure for nonmammalian species.
Andrea Longatti et al.
Nanoscale, 10(29), 14230-14244 (2018-07-17)
Exosomes are extracellular vesicles that mediate cell-to-cell communication by transferring biological cargo, such as DNA, RNA and proteins. Through genetic engineering of exosome-producing cells or manipulation of purified exosomes, it is possible to load exosomes with therapeutic molecules and target
Original Ligand for LTbetaR Is LIGHT: Insight into Evolution of the LT/LTbetaR System
Maeda T, et al.
Journal of Immunology, 201(1), 202-214 (2018)
Fructus Corni extract-induced neuritogenesis in PC12 cells is associated with the suppression of stromal interaction molecule 1 expression and inhibition of Ca2+ influx
Wang X, et al.
Experimental and Therapeutic Medicine, 9(5), 1773-1779 (2015)
Wei Zhang et al.
Cell research, 12(1), 9-18 (2002-04-11)
MAPK families play an important role in complex cellular programs like proliferation, differentiation, development, transformation, and apoptosis. At least three MAPK families have been characterized: extracellular signal-regulated kinase (ERK), Jun kinase (JNK/SAPK) and p38 MAPK. The above effects are fulfilled

Articles

Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.

Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.

Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.

Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.

See All

Protocols

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service