Skip to Content
Merck
All Photos(1)

Documents

F2047

Sigma-Aldrich

Monoclonal Anti-c-Myc−FITC antibody produced in mouse

enhanced validation

clone 9E10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-c-Myc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E10, monoclonal

form

buffered aqueous solution

species reactivity

human

storage condition

protect from light

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

5 μg/mL (suitable for immunoflourescence)

technique(s)

immunocytochemistry: 5.0 μg/mL using mammalian cells transfected with c-Myc tagged fusion protein, fixed with paraformaldehyde.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-c-Myc (mouse IgG1 isotype) is derived from the 9E10 hybridoma, produced by fusion of mouse myeloma cells and splenocytes from BALB/c mice. The antibody is isolated from ascites fluid and conjugated to fluorescein isothiocyanate isomer I (FITC). Monoclonal Anti-c-Myc, FITC conjugate recognizes the c-Myc tag sequence on c-Myc tagged fusion proteins when expressed N- or C-terminal to the fusion protein. c-Myc is a proto-oncogene localized on long arm of human chromosome 8. The encoded protein contains an N-terminal transactivation domain.

Specificity

Monoclonal Anti-c-Myc, FITC conjugate recognizes the c-Myc tag sequence on c-Myc tagged fusion proteins when expressed N- or C-terminal to the fusion protein.

Immunogen

synthetic peptide of the human p62c-Myc protein.

Application

Monoclonal Anti-c-Myc−FITC antibody produced in mouse has been used in immunofluorescence microscopy and immunocytochemistry.

Biochem/physiol Actions

c-Myc plays a vital role in regulation of cell growth, proliferation, differentiation and apoptosis. It is implicated in DNA replication, RNA splicing or transcription. Deregulation in the gene is associated with the development of various types of human cancers. An epitope located within amino acids 410-419, containing the sequence EQKLISEEDL of human c-Myc has been widely used as a tag in many expression vectors, enabling the expression of proteins as c-Myc tag fusion proteins. Epitope tags provide a method to localize gene products in a variety of cell types, to study the topology of proteins and protein complexes, and to identify associated proteins. In addition, it allows characterization of newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.For extended storage, freeze in working aliquots.Repeated freezing and thawing is not recommended.Storage in “frost-free” freezers is not recommended. Ifslight turbidity occurs upon prolonged storage, clarifythe solution by centrifugation before use. Workingdilution samples should be discarded if not used within12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The type III secretion effector NleF of enteropathogenic Escherichia coli activates NF-$\kappa$B early during infection
Pallett MA, et al.
Infection and Immunity, 82(11), 4878-4888 (2014)
Manlio Di Cristina et al.
International journal of cancer, 120(6), 1293-1303 (2006-12-14)
To improve tumor targeting in a subset of patients, where tumor cells do not express the well-known tumor antigens widely used in immunotherapy, we have developed a novel biotechnological tool. It is useful for tumors of various origins for the
Cytoplasmic linker protein CLIP170 negatively regulates TLR4 signaling by targeting the TLR adaptor protein TIRAP
Jakka P, et al.
Journal of immunology (Baltimore, Md. : 1950), 200(2), 704-714 (2018)
The Plasmodium PHIST and RESA-like protein families of human and rodent malaria parasites
Moreira CK, et al.
PLoS ONE, 11(3), e0152510-e0152510 (2016)
Stefano Petrini et al.
Vaccines, 1(4), 463-480 (2013-01-01)
In veterinary medicine, there have been different experiences with the plasmid DNA vaccination. In this area and with the hypothesis to demonstrate the effectiveness of different plasmids encoding porcine respiratory and reproductive syndrome (PRRS), five DNA vaccines against PRRS were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service