Skip to Content
Merck
All Photos(3)

Documents

SAB4501943

Sigma-Aldrich

Anti-MYF5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MYF5, myogenic factor 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 28 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYF5(4617)

General description

Myogenic factor 5 (MYF5) regulates the determination of skeletal myoblasts during embryonic development. Inactivation of MYF5 in mice results in abnormal development of the ribs, which eventually leads to perinatal death. The gene encoding this protein is localized on human chromosome 12q21.

Immunogen

The antiserum was produced against synthesized peptide derived from human MYF5.

Immunogen Range: 21-70

Application

Anti-MYF5 antibody produced in rabbit has been used in western blotting and immunohistochemistry.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of the human myogenic factors 5 and 6 (MYF5, MYF6) gene cluster to 12q21 by in situ hybridization and physical mapping of the locus between D12S350 and D12S106
Cupelli L, et al.
Cell, 72(2-3), 250-251 (1996)
Sonic hedgehog is required for neural crest-dependent patterning of the intrinsic tongue musculature
Shigeru O, et al.
bioRxiv, 154443-154443 (2017)
Myogenic potential of mouse embryonic stem cells lacking functional Pax7 tested in vitro by 5-azacitidine treatment and in vivo in regenerating skeletal muscle
Anita H, et al.
European Journal of Cell Biology, 96(1), 47-60 (2017)
Daniela Calzia et al.
Journal of cellular physiology, 235(4), 3508-3518 (2019-09-25)
Muscle loss is a major problem for many in lifetime. Muscle and bone degeneration has also been observed in individuals exposed to microgravity and in unloading conditions. C2C12 myoblst cells are able to form myotubes, and myofibers and these cells
M A Rudnicki et al.
Cell, 75(7), 1351-1359 (1993-12-31)
Mice carrying null mutations in the myogenic regulatory factors Myf-5 or MyoD have apparently normal skeletal muscle. To address whether these two factors functionally substitute for one another in myogenesis, mice carrying mutant Myf-5 and MyoD genes were interbred. While

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service