Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300069

Sigma-Aldrich

Anti-phospho-IGF1R (pTyr1165/pTyr1166) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CD221 antibody produced in rabbit, Anti-IGFIR antibody produced in rabbit, Anti-JTK13 antibody produced in rabbit, Anti-MGC142170 antibody produced in rabbit, Anti-insulin-like growth factor 1 receptor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~200 kDa
~95 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(T-D-YP-YP-R-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1165/pTyr1166)

Gene Information

human ... IGF1R(3480)

General description

Insulin like growth factor 1 receptor (IGF1R) is encoded by the gene mapped to human chromosome 15q25- qter. The encoded protein is a transmembrane heterotetramer. It is structurally similar to insulin receptor.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 1165/tyrosine 1166 (T-D-Y(p)-Y(p)-R-K), according to the protein IGF1R.

Biochem/physiol Actions

Insulin like growth factor 1 receptor (IGF1R) on binding to its ligand, IGF2 regulates cell proliferation and growth. The encoded protein is also facilitates the normal embryonic and postnatal growth. It plays a crucial role in the embryonic growth. Mutations in IGF2 are associated with Silver–Russell syndrome and ring chromosome 15 syndrome. It is upregulated in ovarian cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

This receptor binds insulin-like growth factor 1 (IGF1) with a high affinity and IGF2 with a lower affinity. It has a tyrosine-protein kinase activity, which is necessary for the activation of the IGF1-stimulated downstream signaling cascade. When present in a hybrid receptor with INSR, binds IGF1.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemizygosity at the insulin-like growth factor I receptor (IGF1R) locus and growth failure in the ring chromosome 15 syndrome.
Peoples R
Cytogenetics and Cell Genetics, 70, 228-234 (1995)
Paternally Inherited IGF2 Mutation and Growth Restriction.
Begemann M
The New England Journal of Medicine, 373, 349-356 (2015)
High IGF2 expression is associated with poor clinical outcome in human ovarian cancer.
Dong Y
Oncology Reports, 34 (2015)
Cell proliferation activities on skin fibroblasts from a short child with absence of one copy of the type 1 insulin-like growth factor receptor (IGF1R) gene and a tall child with three copies of the IGF1R gene.
Okubo Y
The Journal of Clinical Endocrinology and Metabolism, 88, 5981-5988 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service