Skip to Content
Merck
All Photos(3)

Documents

SAB4200576

Sigma-Aldrich

Anti-COX2 antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

human, mouse

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using RAW-264.7 cells stimulated with LPS
western blot: 1.5-3.0 μg/mL using lysates of RAW-264.7 cells stimulated with LPS, and of HEK-293T cells overexpressing human COX2

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COX2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

Cytochrome c oxidase subunit II (COX2) is encoded by the gene mapped to human chromosome 1. The encoded protein is one of the isoforms of COX protein. It is expressed in specific cells such as activated macrophages, monocytes, neutrophils and lymphocytes, in response to inflammatory stimuli such as mitogen, cytokines and growth factors.

Immunogen

synthetic peptide corresponding to an internal sequence of human COX2 (GeneID: 5743), conjugated to KLH. The corresponding sequence has high homology to rat COX2 (83% identity) and to mouse COX2 (82% identity).

Application

Anti-COX2 antibody produced in rabbit has been used in immunohistochemical staining.

Biochem/physiol Actions

Cyclooxygenase (COX) catalyzes the conversion of arachidonic acid (AA) to prostaglandins (PG). Mutation in the gene is associated with the development of gastric cancer. Elevated expression of the gene has been observed in various chronic and autoimmune diseases.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilie Mathieu et al.
Chemical communications (Cambridge, England), 56(57), 7885-7888 (2020-06-11)
A conjugate of a Mn-based superoxide dismutase mimic with a Re-based multimodal probe 1[combining low line] was studied in a cellular model of oxidative stress. Its speciation was investigated using Re and Mn X-fluorescence. Interestingly, 1[combining low line] shows a
Immunohistological study of the effect of extravirgin olive oil on aspartame-treated cerebellum of male albino rat
Baky F A F A .
Menoufia Medical Journal, 29(3), 728-728 (2016)
Detection and Clinical Significance of COX-2 Gene SNPs in Gastric Cancer.
Lu X, et al.
Cell Biochemistry and Biophysics, 72(3), 657-660 (2015)
Hye Suk Baek et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2024-07-31)
This observational study investigated the regulatory mechanism of Pim-1 in inflammatory signaling pathways. THP-1, RAW 264.7, BV2, and Jurkat human T cell lines were used. None. Lipopolysaccharide (LPS) was used to induce inflammation, followed by PIM1 knockdown. Western blot, immunoprecipitation
Daisuke Yokokawa et al.
Experimental and therapeutic medicine, 27(2), 75-75 (2024-01-24)
Cluster of differentiation (CD)44 is a marker of dental pulp stem cells and is involved in odontoblast differentiation and calcification. Chemokine-like receptor 1 (CMKLR1), also known as chemerin receptor 23 (ChemR23) is also expressed in odontoblasts and dental pulp stem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service