Skip to Content
Merck
All Photos(1)

Documents

SAB4200125

Sigma-Aldrich

Anti-Notch3 antibody, Rat monoclonal

purified from hybridoma cell culture

Synonym(s):

Anti-CADASIL, Anti-CASIL, Monoclonal Anti-Notch3 antibody produced in rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

monoclonal

form

buffered aqueous solution

mol wt

antigen ~270 kDa

species reactivity

rat, human

packaging

antibody small pack of 25 μL

technique(s)

western blot: 2-4 μg/mL using using Jurkat cell extracts

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOTCH3(4854)
rat ... Notch3(56761)

General description

Monoclonal Anti-Notch3 (rat IgG2a isotype) is derived from the hybridoma NIZ 8G5 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from the immunized rat.
Members of the Notch gene family encode transmembrane receptors. Mammals have four Notch receptors, Notch1 through Notch4. Notch1 and Notch2 have the highest homology with each other, whereas Notch3 and Notch4 are structurally divergent, lacking the transactivation domain that is found in Notch1 and Notch2. The Notch3 receptor possesses 34 extracellular epidermal growth factor (EGF)-like repeats and three cysteine-rich Notch/LIN12 repeats (LIN). It also contains two protein-protein interaction domains, the RAM domain (R), two nuclear localization signals (NLS), six ankyrin (ANK) repeats and a PEST sequence (P) at its cytoplasmic portion. The NOTCH3 gene is mapped to human chromosome 19p13.12.

Specificity

Monoclonal Anti-Notch3 recognizes human and rat Notch3.

Immunogen

human recombinant full length Notch3.

Application

Monoclonal Anti-Notch3 antibody produced in rat is suitable for immunoblotting.

Biochem/physiol Actions

The Notch receptors are critical for various cell fate decisions. The Notch signaling plays a key role in the normal development of many tissues and cell types. It has diverse effects on differentiation, survival and/or proliferation that are highly dependent on the signal strength and cellular context. Variations in the NOTCH3 gene leads to vascular degenerative disease known as cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL). In addition, upregulation of the NOTCH3 gene indicates active and relapsing human T-cell acute lymphoblastic leukemia (T-ALL).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at -20 °C. For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20 °Cin working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discardedif not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Bellavia et al.
Oncogene, 27(38), 5092-5098 (2008-09-02)
The Notch3 gene was identified, at the beginning of 90s, as the third mammalian Notch and was initially reported as being expressed in proliferating neuroepithelium. Since then, increasing evidence has demonstrated a number of structural and functional differences between Notch3
Freddy Radtke et al.
Nature immunology, 5(3), 247-253 (2004-02-27)
Notch proteins regulate a broad spectrum of cell fate decisions and differentiation processes during fetal and postnatal development. Mammals have four Notch receptors that bind five different ligands. The function of Notch signaling during lymphopoiesis and T cell neoplasia, based
N Turner et al.
Oncogene, 29(14), 2013-2023 (2010-01-27)
Triple negative breast cancers (TNBCs) have a relatively poor prognosis and cannot be effectively treated with current targeted therapies. We searched for genes that have the potential to be therapeutic targets by identifying genes consistently overexpressed when amplified. Fifty-six TNBCs
Silvia Bianchi et al.
Journal of cellular physiology, 207(2), 300-308 (2005-11-05)
Notch proteins encode a family of transmembrane receptors that are part of a signalling transduction system known as Notch signalling, an extremely conserved and widely used mechanism regulating programs governing growth, apoptosis and differentiation in metazoans. Notch signalling begins when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service