Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500350

Sigma-Aldrich

Anti-β-Actin (ACTB) Antibody

chicken polyclonal

Synonym(s):

Anti-β-actin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)

General description

The amino-terminal of actin may function as an antigen. The protein is expressed in the cytoplasm. There are about six isoforms of actin identified.

Immunogen

β-actin antibody was raised against a 16 amino acid peptide from near the carboxy-terminus of human β-actin.

Application

Anti-β-Actin antibody produced in chicken has been used for Western Blotting.

Biochem/physiol Actions

Actin is a cytoskeletal protein that regulates cell motility, secretion, cytoplasmic streaming, phagocytosis and cytokinesis. Mutations in the gene encoding this protein have been associated with Baraitser-Winter cerebrofrontofacial syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Actin amino-acid sequences. Comparison of actins from calf thymus, bovine brain, and SV40-transformed mouse 3T3 cells with rabbit skeletal muscle actin
European Journal of Biochemistry, 90 (1978)
Hye Ji J Kim et al.
Pharmacology research & perspectives, 10(3), e00950-e00950 (2022-04-26)
Sexual dimorphisms are observed in cannabinoid pharmacology. It is widely reported that female animals are more sensitive to the cataleptic, hypothermic, antinociceptive, and anti-locomotive effects of cannabinoid receptor agonists such as CP55,940. Despite awareness of these sex differences, there is
Angela Szeto et al.
Lipids in health and disease, 19(1), 188-188 (2020-08-21)
Obesity and adipose tissue expansion is characterized by a chronic state of systemic inflammation that contributes to disease. The neuropeptide, oxytocin, working through its receptor has been shown to attenuate inflammation in sepsis, wound healing, and cardiovascular disease. The current
?-Actin specifically controls cell growth, migration, and the G-actin pool
Tina M Bunnell
Molecular Biology of the Cell, 22 (2011)
Ling Fu et al.
Oncology letters, 15(4), 5081-5086 (2018-03-20)
Sorafenib is a multikinase inhibitor that is frequently used to treat various types of malignant tumors. However, it has been demonstrated that Sorafenib only has a moderate antitumor efficacy and is associated with numerous side effects in non-small cell lung

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service