Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411195

Sigma-Aldrich

Anti-RLBP1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CRALBP, MGC3663

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 36.5 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RLBP1(6017)

General description

Retinaldehyde binding protein 1 (RLBP1) mapped to human chromosome 15q26, encodes cellular retinaldehyde-binding protein (CRALBP).1 RLBP1 is a 36kDa water soluble protein and is expressed exclusively in retina and pineal gland.
The protein encoded by this gene is a 36kDa water-soluble protein which carries 11-cis-retinaldehyde or 11-cis-retinal as physiologic ligands. It may be a functional component of the visual cycle. Mutations of this gene have been associated with severe rod-cone dystrophy, Bothnia dystrophy (nonsyndromic autosomal recessive retinitis pigmentosa) and retinitis punctata albescens. (provided by RefSeq)

Immunogen

RLBP1 (NP_000317.1, 1 a.a. ~ 317 a.a) full-length human protein.

Sequence
MSEGVGTFRMVPEEEQELRAQLEQLTTKDHGPVFGPCSQLPRHTLQKAKDELNEREETREEAVRELQEMVQAQAASGEELAVAVAERVQEKDSGFFLRFIRARKFNVGRAYELLRGYVNFRLQYPELFDSLSPEAVRCTIEAGYPGVLSSRDKYGRVVMLFNIENWQSQEITFDEILQAYCFILEKLLENEETQINGFCIIENFKGFTMQQAASLRTSDLRKMVDMLQDSFPARFKAIHFIHQPWYFTTTYNVVKPFLKSKLLERVFVHGDDLSGFYQEIDENILPSDFGGTLPKYDGKAVAEQLFGPQAQAENTAF

Biochem/physiol Actions

Retinaldehyde binding protein 1 (RLBP1) plays a vital role in mammalian vision by directing 11-cis-retinoids for the conversion of photobleached opsin molecules into photosensitive visual pigments. Genetic variations in the gene leads to various autosomal recessive progressive retinopathy, including Bothnia dystrophy (BD) and retinitis, punctata albescens (RPA), Newfoundland rod-cone dystrophy (NFRCD), retinitis pigmentosa (RP) and fundus albipunctatus (FA).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elodie Dessalces et al.
JAMA ophthalmology, 131(10), 1314-1323 (2013-08-10)
Retinitis punctata albescens (RPA) is an autosomal recessive form of retinitis pigmentosa characterized by white dotlike deposits in the fundus, in most cases caused by mutations in RLBP1. To study disease progression and visual function in RPA. We performed clinical
Xiaoqin He et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(44), 18545-18550 (2009-10-23)
Cellular retinaldehyde-binding protein (CRALBP) is essential for mammalian vision by routing 11-cis-retinoids for the conversion of photobleached opsin molecules into photosensitive visual pigments. The arginine-to-tryptophan missense mutation in position 234 (R234W) in the human gene RLBP1 encoding CRALBP compromises visual
Stephanie Hipp et al.
Acta ophthalmologica, 93(4), e281-e286 (2014-11-29)
Mutations in the RLBP1 gene encoding the cellular retinaldehyde-binding protein (CRALBP) cause autosomal recessive progressive retinopathy, such as retinitis punctata albescens (RPA), Bothnia-type dystrophy (BD), Newfoundland rod-cone dystrophy (NFRCD), retinitis pigmentosa (RP) and fundus albipunctatus (FA). We present the clinical
Molecular clues to Bothnia-type retinal dystrophy.
Xiaoqin He et al.
Advances in experimental medicine and biology, 723, 589-594 (2011-12-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service