Skip to Content
Merck
All Photos(1)

Key Documents

S9317

Sigma-Aldrich

PAR-2 (1-6) amide (mouse, rat) trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

PAR2-AP, SLIGRL−NH2, Ser-Leu-Ile-Gly-Arg-Leu-amide trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H56O7N10 · C2HF3O2
Molecular Weight:
770.84
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](N)CO)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(N)=O

InChI

1S/C29H56N10O7/c1-7-17(6)23(39-27(45)21(12-16(4)5)38-25(43)18(30)14-40)28(46)35-13-22(41)36-19(9-8-10-34-29(32)33)26(44)37-20(24(31)42)11-15(2)3/h15-21,23,40H,7-14,30H2,1-6H3,(H2,31,42)(H,35,46)(H,36,41)(H,37,44)(H,38,43)(H,39,45)(H4,32,33,34)/t17-,18-,19-,20-,21-,23-/m0/s1

InChI key

SGPMJRPYYIJZPC-JYAZKYGWSA-N

Amino Acid Sequence

Ser-Leu-Ile-Gly-Arg-Leu-NH2

Application

PAR-2 (1-6) amide (mouse, rat) trifluoroacetate salt has been used as a proteinase-activated receptor 2 (PAR-2) agonist to induce the expression of Hh and Hippo genes in Hek293A cells.

Biochem/physiol Actions

Selective proteinase-activated receptor 2 (PAR2) peptide agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Yes-associated protein controls the cell density regulation of Hedgehog signaling
M. Tariki
Oncogenesis (2014)
William G Darby et al.
Pharmacological research, 133, 152-159 (2018-05-23)
The aim of this study was to better understand the role of TRPV4 in the regulation of blood vessel dilatation by blood flow and activation of GPCRs. Using pressure myography, the dilator responses to the TRPV4 agonist GSK1016790A and to
Imen Nasri et al.
Pharmaceutical biology, 55(1), 124-131 (2016-12-08)
Diplotaxis harra (Forssk.) Boiss. (Brassicaceae) is traditionally used as an antidiabetic, anti-inflammatory or anticancer agent. In these pathologies, the glycogen synthase kinase 3 β (GSK3β) is overactivated and represents an interesting therapeutic target. Several flavonoids can inhibit GSK3β and the
Michael R Sutherland et al.
Journal of thrombosis and haemostasis : JTH, 17(3), 482-491 (2019-01-20)
Essentials The coagulation initiator, tissue factor (TF), is on the herpes simplex virus 1 (HSV1) surface. HSV1 surface TF was examined in mice as an antiviral target since it enhances infection in vitro. HSV1 surface TF facilitated infection of all organs
Timo Buhl et al.
Frontiers in immunology, 11, 1740-1740 (2020-09-10)
Background: Activation of protease-activated receptor-2 (PAR2) has been implicated in inflammation, pruritus, and skin barrier regulation, all characteristics of atopic dermatitis (AD), as well as Netherton syndrome which has similar characteristics. However, understanding the precise role of PAR2 on neuro-immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service