Skip to Content
Merck
All Photos(1)

Key Documents

S9263

Sigma-Aldrich

Swainsonine

solid, ≥99% (HPLC)

Synonym(s):

(1S,2R,8R,8aR)-1,2,8-Octahydroindolizidinetriol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
Beilstein:
4175740
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

Swainsonine, synthetic

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

solubility

methanol: 5 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

O[C@@H]1CCCN2C[C@@H](O)[C@@H](O)C12

InChI

1S/C8H15NO3/c10-5-2-1-3-9-4-6(11)8(12)7(5)9/h5-8,10-12H,1-4H2/t5-,6-,7?,8-/m1/s1

InChI key

FXUAIOOAOAVCGD-DCDLSZRSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Swainsonine is produced by endophytes, plant and insect pathogens. It is synthesized from the pipecolic acid and mevalonic acid.[1] Swainsonine is a water-soluble indole alkaloid.[2]

Application

Swainsonine has been used:
  • to test its effect on neural stem cell proliferation[2]
  • to inhibit α-mannosidase in TC-1 tumor cells[3]
  • to inhibit β1,6-branching synthesis in hepatocellular carcinoma (HCC) cells[4]

Swainsonine is a potent inhibitor of various α-mannosidases, especially of α-mannosidase II. It inhibits glycoprotein processing and also acts as immune modulator.

Biochem/physiol Actions

Swainsonine is an inhibitor of α-mannosidase in endoplasmic reticulum and Golgi apparatus. It also activates the natural killer cells.[3] Swainsonine elicits neurologic toxicity and its exposure impairs learning and memory resulting in cognitive deficit.[2] It inhibits hepatocellular carcinoma and sensitizes them to respond to anti-cancer drug paclitaxel.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Swainsonine, an alpha-mannosidase inhibitor, may worsen cervical cancer progression through the increase in myeloid derived suppressor cells population
SilveiraCRF, et al.
PLoS ONE, 14(3), e0213184-e0213184 (2019)
Exposure to swainsonine impairs adult neurogenesis and spatial learning and memory
Wang J, et al.
Toxicology Letters, 232(1), 263-270 (2015)
N-glycosylation by N-acetylglucosaminyltransferase V enhances the interaction of CD147/basigin with integrin beta1 and promotes HCC metastasis
Cui J, et al.
The Journal of Pathology, 245(1), 41-52 (2018)
Swainsonine biosynthesis genes in diverse symbiotic and pathogenic fungi
Cook D, et al.
G3 (Bethesda, Md.), 7(6), 1791-1797 (2017)
Caio Raony Farina Silveira et al.
PloS one, 14(3), e0213184-e0213184 (2019-03-07)
Cervical cancer, caused by high oncogenic risk Human Papillomavirus (HPV) infection, continues to be a public health problem, mainly in developing countries. Using peptide phage display as a tool to identify potential molecular targets in HPV associated tumors, we identified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service