Skip to Content
Merck
All Photos(1)

Key Documents

V0882

Sigma-Aldrich

D-Val-Leu-Lys 4-nitroanilide dihydrochloride

plasmin substrate

Synonym(s):

D-Val-Leu-Lys-pNA dihydrochloride, D-Valyl-L-leucyl-L-lysine 4-nitroanilide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H38N6O5 · 2HCl
CAS Number:
Molecular Weight:
551.51
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to yellow

storage temp.

−20°C

SMILES string

Cl.Cl.CC(C)C[C@H](NC(=O)[C@H](N)C(C)C)C(=O)N[C@@H](CCCCN)C(=O)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C23H38N6O5.2ClH/c1-14(2)13-19(28-23(32)20(25)15(3)4)22(31)27-18(7-5-6-12-24)21(30)26-16-8-10-17(11-9-16)29(33)34;;/h8-11,14-15,18-20H,5-7,12-13,24-25H2,1-4H3,(H,26,30)(H,27,31)(H,28,32);2*1H/t18-,19-,20+;;/m0../s1

InChI key

VESQMNNSPPEOSZ-ZLARAOTRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Val-Leu-Lys 4-nitroanilide dihydrochloride has been used as a chromogenic substrate to determine the formation of plasmin from plasminogen in amidolytic activity assay[1] and plasminogen activating assay.[2]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ueshima et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 7(5), 522-529 (1996-07-01)
Staphylokinase (SAK), produced by Staphylococcus aureus, induces fibrinolytic activity in circulation without systemic fibrinolytic activation. Since the effect of blood vessels on the activity of SAK has not yet been clarified, plasminogen activator (PA) activity of SAK in the presence
M Ieko et al.
Seminars in thrombosis and hemostasis, 26(1), 85-90 (2000-05-11)
Antiphospholipid antibodies (aPLs) are associated with an increased incidence of thrombosis, but the mechanisms responsible for thrombosis are unclear. The present study investigated the effect of both beta2-glycoprotein I (beta2-GPI) and aPLs on the activity of extrinsic fibrinolysis. The remaining
J F Liang et al.
Thrombosis research, 97(5), 349-358 (2000-03-10)
The effects and possible role of heparin on tissue plasminogen activator-mediated plasminogen activation was thoroughly investigated. Direct analysis by sodium dodecyl sulfate-polyacrylamide gel electrophoresis demonstrated that heparin increased the conversion of plasminogen to plasmin. Experiments by fluorescence quenching suggested that
K Kolev et al.
Biochimica et biophysica acta, 1247(2), 239-245 (1995-03-15)
The effect of plasmin substrates D-valyl-L-leucyl-lysine-p-nitroanilide (S-2251) and H-D-norleucyl-hexahydrotyrosyl-lysine-p-nitro-anilide (Spectrozyme-PL) on the rate of activation of native human plasminogen in physiological salt solution is studied. Plasminogen activation by two-chain urokinase-type plasminogen activator (urokinase), two-chain tissue-type plasminogen activator (tc-tPA) or trypsin
Genetic modification of the streptokinase gene to study the interaction of streptokinase with plasminogen.
S Lizano et al.
Developments in biological standardization, 85, 195-198 (1995-01-01)

Protocols

Enzymatic Assay of Plasmin with D-Val-Leu-Lys-p-Nitroanilide Dihydrochloride

Protocol for Enzymatic Assay of Plasmin with D-Val-Leu-Lys-p-Nitroanilide Dihydrochloride

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service