Skip to Content
Merck
All Photos(1)

Key Documents

S5143

Sigma-Aldrich

Serum minus IgG human

lyophilized powder

Synonym(s):

Human Serum without IgG, IgG-Free Human Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

human

Quality Level

conjugate

unconjugated

form

lyophilized powder

contains

15 mM sodium azide

packaging

vial of 0.5 mL

storage temp.

2-8°C

General description

This product is pooled, lyophilized human serum deficient of IgG.

Application

Serum minus IgG human has been used in:
  • immunoelectrophoresis
  • immunodiffusion
  • enzyme-linked immunosorbent assay (ELISA)
  • serum analysis
  • serum analysis by MISPE−PE−MS and molecularly imprinted solid phase extraction (MISPE)−pulsed elution (PE) method coupled with electrospray mass spectrometry (MS)

Serum minus IgG human was used for screening of cephalexin in human plasma and serum by molecularly imprinted solid phase extraction technique.

Quantity

Each vial contains at least 25 mg protein

Reconstitution

Reconstitute with 0.5 mL deionized water

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecularly imprinted solid phase extraction for rapid screening of cephalexin in human plasma and serum
Lai EPC and Wu SG
Analytica Chimica Acta, 481(2), 165-174 (2003)
The Immunoglobulins: Structure and Function (1998)
Stanley G Wu et al.
Journal of pharmaceutical and biomedical analysis, 36(3), 483-490 (2004-11-04)
A highly selective molecularly imprinted solid phase extraction (MISPE)-pulsed elution (PE) method coupled with electrospray mass spectrometry (MS) was developed for the rapid screening and determination of cephalexin in alpha-aminocephalosporin antibiotics. This method involved the solid phase extraction of cephalexin
Molecular Genetics of Immunoglobulin (1987)
Molecularly imprinted solid phase extraction-pulsed elution-mass spectrometry for determination of cephalexin and alpha-aminocephalosporin antibiotics in human serum
Wu SG, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(3), 483-490 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service