Skip to Content
Merck
All Photos(4)

Documents

S4140

Sigma-Aldrich

D-Saccharic acid potassium salt

≥98% (GC)

Synonym(s):

Glucaric Acid, Glucosaccharic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9KO8
CAS Number:
Molecular Weight:
248.23
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (GC)

form

powder

technique(s)

gas chromatography (GC): suitable

color

white to off-white

mp

188  °C ((370 °F ))

solubility

soluble, clear to very slightly hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

[K+].O[C@H]([C@@H](O)[C@@H](O)C([O-])=O)[C@H](O)C(O)=O

InChI

1S/C6H10O8.K/c7-1(3(9)5(11)12)2(8)4(10)6(13)14;/h1-4,7-10H,(H,11,12)(H,13,14);/q;+1/p-1/t1-,2-,3-,4+;/m1./s1

InChI key

UBYZGUWQNIEQMH-IAMDKXPVSA-M

Looking for similar products? Visit Product Comparison Guide

General description

D-Saccharic acid potassium salt is present in plants and animals. It is produced by the chemical oxidation of glucose with nitric acid. D-Saccharic acid lowers cholesterol and has chemotherapeutic property. It also has antimutagenic activity.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of glucaric acid from a synthetic pathway in recombinant Escherichia coli
Moon TS, et al.
Applied and Environmental Microbiology, 75(3), 589-595 (2009)
N Yoshimi et al.
International journal of oncology, 16(1), 43-48 (1999-12-22)
While calcium D-glucarate was shown to inhibit chemical carcinogenesis in various animal models, the effect of potassium hydrogen D-glucarate has not been extensively investigated. In the present study, potassium hydrogen D-glucarate markedly inhibited azoxymethane (AOM)-induced colon carcinogenesis in male F344
Essential Oil Safety - E-Book: A Guide for Health Care Professionals (2013)
Ling Li et al.
Molecular cell, 80(3), 525-540 (2020-10-18)
Well-balanced and timed metabolism is essential for making a high-quality egg. However, the metabolic framework that supports oocyte development remains poorly understood. Here, we obtained the temporal metabolome profiles of mouse oocytes during in vivo maturation by isolating large number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service