Skip to Content
Merck
All Photos(4)

Documents

S0375

Sigma-Aldrich

D-Saccharic acid 1,4-lactone monohydrate

≥98.0% (HPLC)

Synonym(s):

D-Glucaric acid-1,4-lactone, D-Saccharolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O7 · H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
85594
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (HPLC)

form

powder

impurities

≤10.0% water (Karl Fischer)

color

white

mp

91 - 93 °C ((196 - 199 °F ))

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

O.O[C@@H]1[C@@H](O)C(=O)O[C@@H]1[C@H](O)C(O)=O

InChI

1S/C6H8O7.H2O/c7-1-2(8)6(12)13-4(1)3(9)5(10)11;/h1-4,7-9H,(H,10,11);1H2/t1-,2-,3+,4+;/m1./s1

InChI key

NPFKVZHSFSFLPU-QGBSHYGGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Saccharic acid 1,4-lactone monohydrate (DSAL) has an ability to inhibit the activity of β-glucuronidase enzyme.It also potentially inhibits Klotho deglycosylation of excitatory amino acid transporters (EAATs) and peptide transporters (PEPTs).

Application

D-Saccharic acid 1,4-lactone monohydrate has been used as a constituent of phosphate buffer to block incidental β-glucuronidase activity in Streptococcus pneumoniae hyaluronan lyase (SpnHL) activity assay. It has also been used as a component of reaction mixture in Iduronidase activity assay and UDP-glucuronosyltransferase 1 (UGT1A1) inhibition assay.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cas9-mediated allelic exchange repairs compound heterozygous recessive mutations in mice
Wang D, et al.
Nature Biotechnology, 36(9), 839-839 (2018)
In vitro OATP1B1 and OATP1B3 inhibition is associated with observations of benign clinical unconjugated hyperbilirubinemia
Chiou W, et al.
Xenobiotica, 44(3), 276-282 (2014)
Klotho preservation by Rhein promotes toll-like receptor 4 proteolysis and attenuates lipopolysaccharide-induced acute kidney injury
Bi F, et al.
Journal of Molecular Medicine, 96(9), 915-927 (2018)
Magnetic macroporous bead cellulose functionalised with recombinant hyaluronan lyase for controllable hyaluronan fragmentation
Kavsparova J, et al.
Process Biochemistry (Oxford, United Kingdom), 72, 105-111 (2018)
Gertrud E Morlock et al.
Antioxidants (Basel, Switzerland), 10(1) (2021-01-21)
The antioxidative activity of Camelia sinensis tea and especially powdered tea extracts on the market, among others used as added value in functional foods, can considerably vary due to not only natural variance, but also adulteration and falsification. Thus, an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service