Skip to Content
Merck
All Photos(2)

Documents

S3378

Sigma-Aldrich

Spironolactone

97.0-103.0% (HPLC), powder, aldosterone receptor antagonist

Synonym(s):

4-Pregnen-21-oic acid-17α-ol-3-one-7α-thiol γ-lactone 7-acetate, 7α-(Acetylthio)-17α-hydroxy-3-oxopregn-4-ene-21-carboxylic acid γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O4S
CAS Number:
Molecular Weight:
416.57
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

Spironolactone, 97.0-103.0%

Quality Level

Assay

97.0-103.0%

mp

207-208 °C (lit.)

solubility

chloroform: complete 50 mg/ml, clear, faintly yellow

SMILES string

CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]45CCC(=O)O5)[C@H]13

InChI

1S/C24H32O4S/c1-14(25)29-19-13-15-12-16(26)4-8-22(15,2)17-5-9-23(3)18(21(17)19)6-10-24(23)11-7-20(27)28-24/h12,17-19,21H,4-11,13H2,1-3H3/t17-,18-,19+,21+,22-,23-,24+/m0/s1

InChI key

LXMSZDCAJNLERA-ZHYRCANASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Spironolactone was added to rat diet to study the effect of long-term spironolactone use on renal function.

Biochem/physiol Actions

Spironolactone reduces aldosterone-induced potassium/magnesium loss and myocardial fibrosis. It reduces hypertension, improves the endothelial function and reduces the overall morbidity and mortality in patients with chronic heart failure. Spironolactone improves nitric oxide bioactivity and vascular endothelial vasodilator dysfunction.
Spironolactone is a competitive aldosterone receptor antagonist. Used as potassium sparing diuretic.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Spironolactone yields clear, faint yellow solution in chloroform at 50 mg/ml.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sato et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 22(1), 17-22 (1999-04-30)
There is increasing evidence for important cardiovascular effects of aldosterone via classical mineralocorticoid receptors in the heart. Administration of aldosterone with excess salt produces both cardiac hypertrophy and interstitial cardiac fibrosis in rats, and concomitant administration of potassium canrenoate at
Femke Waanders et al.
American journal of physiology. Renal physiology, 296(5), F1072-F1079 (2009-02-27)
Chronic transplant dysfunction (CTD) is the leading cause of long-term renal allograft loss and is characterized by specific histological lesions including transplant vasculopathy, interstitial fibrosis, and focal glomerulosclerosis. Increasing evidence indicates that aldosterone is a direct mediator of renal damage
P Moghetti et al.
The Journal of clinical endocrinology and metabolism, 84(4), 1250-1254 (1999-04-13)
GnRH agonists (GnRHa) have recently been proposed for the treatment of hirsutism in women with the polycystic ovary syndrome (PCOS). As most of these subjects have increased androgen secretion from both ovaries and adrenal glands, the association of GnRHa with
Donna A Volpe et al.
The AAPS journal, 16(1), 172-180 (2013-12-18)
Drug interactions due to efflux transporters may result in one drug increasing or decreasing the systemic exposure of a second drug. The potential for in vivo drug interactions is estimated through in vitro cell assays. Variability in in vitro parameter
Jakob Nielsen et al.
American journal of physiology. Renal physiology, 283(5), F923-F933 (2002-10-10)
Renal tubule profiling studies were carried out to investigate the long-term effects of administration of spironolactone, a mineralocorticoid receptor antagonist, on abundances of the major Na transporter and Na channel proteins along the rat renal tubule. Oral administration of spironolactone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service