Skip to Content
Merck
All Photos(1)

Key Documents

E6657

Sigma-Aldrich

Eplerenone

≥98% (HPLC)

Synonym(s):

9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic acid, gamma-lactone, methyl ester, Cgp 30083, Eplerenone [USAN], Epoxymexrenone, HSDB 7522, Inspra, Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, gamma-lactone, methyl ester, (7alpha,11alpha,17alpha)-, SC-66110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O6
CAS Number:
Molecular Weight:
414.49
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)
H2O: insoluble

storage temp.

2-8°C

SMILES string

O1[C@]32[C@H]1C[C@@]4([C@@]6(OC(=O)CC6)CC[C@H]4[C@@H]3[C@@H](CC5=CC(=O)CC[C@@]52C)C(=O)OC)C

InChI

1S/C24H30O6/c1-21-7-4-14(25)10-13(21)11-15(20(27)28-3)19-16-5-8-23(9-6-18(26)30-23)22(16,2)12-17-24(19,21)29-17/h10,15-17,19H,4-9,11-12H2,1-3H3/t15-,16+,17-,19+,21+,22+,23-,24-/m1/s1

InChI key

JUKPWJGBANNWMW-VWBFHTRKSA-N

Gene Information

human ... NR3C2(4306)

Looking for similar products? Visit Product Comparison Guide

General description

Eplerenone is an aldosterone blocker, which particularly blocks mineralocorticoid receptor. It reduces cardiovascular risk in patients following acute myocardial infarction. Eplerenone is used for treating hypertension and central serous retinopathy. It exhibits potassium sparing diuretic effect.

Application

Eplerenone has been used in quantitative real time (RT-qPCR) or western blotting assays.

Biochem/physiol Actions

Eplerenone is an aldosterone antagonist more specific for the mineralocorticoid receptor than spironolactone (S3378), having lower affinity for progesterone, androgen, and glucocorticoid receptors.
Eplerenone is an aldosterone antagonist.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Mondadori et al.
Psychopharmacology, 124(4), 380-383 (1996-04-01)
The retention performance of mice in a passive-avoidance task was facilitated by low doses (0.3 mg/kg) of the competitive NMDA-receptor blocker CGP 37849, but impaired by high doses (30 mg/kg). The facilitatory effect was selectively suppressed by elevation of the
Tracy Ann Williams et al.
The Journal of clinical endocrinology and metabolism, 100(9), E1262-E1269 (2015-07-16)
Coexisting prolactinoma-primary aldosteronism (PA) is infrequently reported. The objective of the study was to identify patients with prolactinoma-PA and test the hypothesis that elevated prolactin (PRL) concentrations play a role in PA pathogenesis. Hyperprolactinemia/prolactinoma was diagnosed in PA patients from
Eplerenone survival benefits in heart failure patients post-myocardial infarction are independent from its diuretic and potassium-sparing effects: insights from an EPHESUS (Eplerenone Post-Acute Myocardial Infarction Heart Failure Efficacy and Survival Study) substudy
Rossignol P, et al.
Journal of the American College of Cardiology, 58(19), 1958-1966 (2011)
Oral eplerenone for treatment of chronic central serous chorioretinopathy: a case series
Sal DA, et al.
Ophthalmic Surgery, Lasers & Imaging Retina, 46(4), 439-444 (2015)
M de Gasparo et al.
Journal of steroid biochemistry, 32(1B), 223-227 (1989-01-01)
The use of spironolactone in the treatment of hypertension has been limited by the occurrence of sexual side effects, mainly menstrual disturbances in women and gynaecomastia in men. In order to minimize this limitation on the use of an effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service