Skip to Content
Merck
All Photos(2)

Key Documents

RES0114M-A7

SAFC

MES sodium salt

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid sodium salt, 4-Morpholineethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NNaO4S
CAS Number:
Molecular Weight:
217.22
Beilstein:
3765682
EC Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial, tested

useful pH range

5.5-6.7

pKa 

6.1

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

[Na+].[O-]S(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.Na/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IRHWMYKYLWNHTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

MES Sodium is a biological buffer often referred to as a “Good′s” buffer. The pKa of MES is 5.96 which makes MES an ideal candidate for cell culture media and protein based buffer formulations to maintain a stable environment in solution. MES Sodium is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

MES Sodium is used in cell culture media, biopharmaceutical buffer formulations both upstream and downstream and diagnostic reagents. MES based buffers are used in purification bioprocesses of antibodies, peptides, proteins and blood components.

Packaging

Product is available in the following package sizes:
RES0114M-A701X: 100g container
RES0114M-A702X 1kg container
RES0114M-A704X: 10kg container
RES0114M-A705X: 25kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pasi S Vuorinen et al.
Journal of chromatography. A, 990(1-2), 45-52 (2003-04-11)
A contactless conductivity detector integrated into the capillary cassette of Agilent (3D)CE equipment is described. The detector is user-friendly, compact and easily modified. The UV detector of the (3D)CE equipment is available parallel with the contactless conductivity detector increasing the
Christian Engelbrekt et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(13), 2844-2853 (2010-08-18)
Monodisperse platinum nanoparticles (PtNPs) were synthesized by a green recipe. Glucose serves as a reducing agent and starch as a stabilization agent to protect the freshly formed PtNP cores in buffered aqueous solutions. Among the ten buffers studied, 2-(N-morpholino)ethanesulfonic acid
Kefang Xie et al.
Acta crystallographica. Section D, Biological crystallography, 61(Pt 10), 1386-1394 (2005-10-06)
The NNA7 Fab antibody fragment recognizes the human N-type blood-group antigen comprised of the N-terminal glycopeptide of glycophorin A (GPA). A mutant form of this Fab fragment, NNA7-G91S, exhibits markedly reduced antigen binding. To provide insight into how these Fab
Ekta G Ahuja et al.
Acta crystallographica. Section D, Biological crystallography, 60(Pt 6), 1129-1131 (2004-05-26)
Phenazines are broad-spectrum antibiotic metabolites produced by organisms such as Pseudomonas and Streptomyces. Phenazines have been shown to enhance microbial competitiveness and the pathogenic potential of the organisms that synthesize them. PhzA (163 residues, approximate molecular weight 18.7 kDa) is
Elisabet Fuguet et al.
Electrophoresis, 29(13), 2841-2851 (2008-06-12)
The performance of the most common and also some other less common CE buffers has been tested for the pKa determination of several types of compounds (pyridine, amines, and phenols). The selected buffers cover a pH ranging from 3.7 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service