Skip to Content
Merck
All Photos(1)

Key Documents

RAB0849

Sigma-Aldrich

Mouse Tnfsf10 / Tumor Necrosis Factor Ligand Superfamily Member 10 ELISA Kit

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 2.46-600 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Tnfsf10(22035)

General description

The antibody pair provided in this kit recognizes mouse TRAIL/TNFSF10.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) activates the extrinsic apoptotic pathway by binding death receptors (DR)-4 and 5. In addition to DR 4 and 5, TRAIL activates several other receptors such as decoy receptor (DcR)1, DcR2 and a soluble receptor called osteoprotegerin. TRAIL plays a vital role in the inhibition of autoimmune inflammation and cell cycle progression. TRAIL selectively kills tumor cells without damaging normal tissues and therefore, it might act as a tumor-selective anticancer agent. TRAIL-induced apoptosis is involved in various cellular processes including activation-induced cell death, homeostasis, intrathymic negative selection, autoimmunity suppression, and immune surveillance. Repeated dosage of recombinant soluble TRAIL triggers apoptosis, inhibits tumor progression, and increases viability of mice suffering from cancer.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of soluble TRAIL induces apoptosis in human lung adenocarcinoma and inhibits growth of tumor xenografts in nude mice
Shi J
Cancer Research, 65 (2005)
Exosomes secreted by human placenta carry functional Fas ligand and TRAIL molecules and convey apoptosis in activated immune cells, suggesting exosome-mediated immune privilege of the fetus
Stenqvist AC
Journal of Immunology, 191, 5515-5523 (2013)
Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) induced mitochondrial pathway to apoptosis and caspase activation is potentiated by phospholipid scramblase-3
Ndebele K
Apoptosis, 13(7) (2008)
Lin Li et al.
Acta biochimica et biophysica Sinica, 46(6), 471-476 (2014-04-18)
The apoptotic ligand TNF-related apoptosis-inducing ligand (TRAIL) is believed to be a promising candidate for cancer gene therapy, yet gene therapy strategies to tackle this disease systemically are often impaired by inefficient delivery of the vector to the tumor tissue.
A Ray et al.
Leukemia, 28(8), 1716-1724 (2014-01-31)
Our prior study in multiple myeloma (MM) patients showed increased numbers of plasmacytoid dendritic cells (pDCs) in the bone marrow (BM), which both contribute to immune dysfunction as well as promote tumor cell growth, survival and drug resistance. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service