RAB0369
Human MMP-7 ELISA Kit
on for cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 150 pg/mL
standard curve range: 0.137-100 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... MMP7(4316)
General description
The human MMP-7 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MMP-7 in cell culture supernatants. Matrix metalloproteinase-7 (MMP-7)/matrilysin is a small proteolytic enzyme. It secretes zinc- and calcium-dependent endopeptidase. It is expressed in normal endometrial, bronchial, ductal, skin glandular epithelial, urogenital, and gastrointestinal tissues. MMP-7 plays a key role in cell proliferation, migration, invasion, and apoptosis. Elevated expression of MMP-7 is seen in several cancers, such as esophageal, gastric, gallbladder, pancreatic, colorectal, bladder, ovarian, breast, lung, and melanoma.
Immunogen
Recombinant Human MMP7
Application
Human MMP-7 ELISA Kit has been used in ELISA to measure the recovery of the cancer biomarkers/MMP-7 in isothermally vitrified samples. It has also been used in ELISA to determine MMP7 concentrations in multiple myeloma (MM) cell culture supernatants.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hai-Yang Liao et al.
Clinical biochemistry, 92, 9-18 (2021-03-14)
Matrix metalloproteinase-7 (MMP-7) is a small proteolytic enzyme that secretes zinc and calcium endopeptidases. It can degrade a variety of extracellular matrix substrates and other substrates and plays important regulatory roles in many human pathophysiological processes. Since its discovery, MMP-7
Guobao Wang et al.
Arthritis & rheumatology (Hoboken, N.J.), 73(2), 265-275 (2020-09-07)
Flares of lupus nephritis (LN) are frequent and associated with impaired renal prognosis. One major management obstacle in LN flare is the lack of effective methods to identify at-risk patients earlier in their disease course. This study was undertaken to
Anita M Tuomainen et al.
European journal of preventive cardiology, 21(7), 806-812 (2012-10-20)
Extracellular matrix-degrading proteinases are upregulated in atherosclerotic lesions and can contribute to subsequent pathological events. In the present nested case-control study, we investigated the association of serum concentrations of matrix metalloproteinases MMP-7, MMP-8, and MMP-13, tissue inhibitor of metalloproteinase-1 (TIMP-1)
Vladimir Riabov et al.
Acta biomaterialia, 53, 389-398 (2017-02-06)
The immediate tissue microenvironment of implanted biomedical devices and engineered tissues is highly influential on their long term fate and efficacy. The creation of a long-term anti-inflammatory microenvironment around implants and artificial tissues can facilitate their integration. Macrophages are highly
Kasturi Chatterjee et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 32(8), 4560-4572 (2018-03-21)
Endometriosis, characterized by extrauterine development of endometrial glands and stroma, is associated with increased risk of ovarian cancer development. In the present study, we investigated the role of matrix metalloproteinase-7 (MMP-7) on epithelial-mesenchymal transition (EMT) during ovarian endometriosis ( N
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service