Skip to Content
Merck
All Photos(2)

Key Documents

RAB0286

Sigma-Aldrich

Human IL-2 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

IL-1 beta, Interleukin-1 beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type urine
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 2.07-1500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL2(3558)

General description

The Human IL-2 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-2 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-2

Application

Human IL-2 ELISA Kit has been used in the analysis of interleukins levels in human plasma.[1]

Biochem/physiol Actions

IL-2 (interleukin 2) is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation.[2] It is a type I cytokine.[3] IL-2/IL-2R (interleukin 2/IL-2 receptor) signaling is required for T-cell proliferation and other fundamental functions which are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK (natural killer) cells, lymphokine activated killer cells, monocytes, macrophages and oligodendrocytes.[2][4] It is associated with recurrent oral ulcer, asthma and Mycoplasma pneumoniae infection in children.[5][6] Absence of IL2 in mice causes fatal lymphoproliferative inflammatory disease (IL-2 deficiency syndrome).[7] IL-2 signaling involves three pathways: phosphoinositol 3-kinase (PI3-K)/AKT, Ras-MAPK (mitogen activated protein kinase), and JAK-STAT (Janus kinase - signal transducers and activators of transcription) pathways.[3]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BbvI & RsaI Restriction of AR-DBD, IL-6, IL-8, and SPF Discriminate Prostate Cancer
Hessien M, et al.
Biomedica : Revista del Instituto Nacional de Salud, 34(4), 236-236 (2018)
Ruihong Lan et al.
Experimental and therapeutic medicine, 22(4), 1055-1055 (2021-08-27)
Placental trophoblast apoptosis is a major pathological feature of preeclampsia. Fas has been reported to be highly expressed in the placentas of patients with preeclampsia. However, the role and underlying mechanisms of Fas in the pathogenesis of preeclampsia have not
IL-2 family cytokines: new insights into the complex roles of IL-2 as a broad regulator of T helper cell differentiation.
Liao W
Current Opinion in Immunology, 23, 598-604 (2011)
Homeostatic maintenance of natural Foxp3(+) CD25(+) CD4(+) regulatory T cells by interleukin (IL)-2 and induction of autoimmune disease by IL-2 neutralization.
Setoguchi R
The Journal of Experimental Medicine, 201, 723-735 (2005)
The interleukin (IL) 2 receptor beta chain is shared by IL-2 and a cytokine, provisionally designated IL-T, that stimulates T-cell proliferation and the induction of lymphokine-activated killer cells.
Bamford RN
Proceedings of the National Academy of Sciences of the USA, 91, 4940-4944 (1994)

Articles

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service