EZHTNFA-150K
Human TNF-α ELISA Kit
detects TNF-α in 50 μL serum, plasma or cell culture supernatent
Synonym(s):
Human TNF-A ELISA, TNF-A ELISA, TNF-Alpha, Tumour Necrosis Factor Alpha ELISA
About This Item
Recommended Products
Product Name
Human TNF-α Conferma® ELISA Kit, kit sufficient for 96 wells
Quality Level
species reactivity
human
packaging
kit of 1 × 96 wells
parameter
50 μL sample volume (4hr assay)
assay range
linearity: 89-113%
(Serum, 101%)
linearity: 98-110%
(Plasma, 103%)
recovery range: 81-111%
(Plasma, 97%)
recovery range: 85-105%
(Serum,95%)
inter-assay cv: ≤7%
intra-assay cv: ≤3%
sensitivity: 1.65 pg/mL
standard curve range: 1.65-400 pg/mL
technique(s)
ELISA: suitable
input
sample type cell culture supernatant
sample type serum
sample type plasma (K2 EDTA)
application(s)
research use
detection method
colorimetric (450nm/590nm)
shipped in
wet ice
storage temp.
2-8°C
Related Categories
General description
Research Category: Inflammation
Research Sub-Category:
Alzheimer′s Disease
Aging
Inflammation
Neurodegenerative Diseases
Metabolic Disease
Sepsis Viral Diseases
Cardiovascular Disease
Specificity
Application
Features and Benefits
Legal Information
Disclaimer
recommended
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn about the Conferma® ELISA development and manufacturing methods that provide strong sample detection and long-term assay and lot consistency, giving you confidence in your research.
Learn about the Conferma® ELISA development and manufacturing methods that provide strong sample detection and long-term assay and lot consistency, giving you confidence in your research.
Learn about the Conferma® ELISA development and manufacturing methods that provide strong sample detection and long-term assay and lot consistency, giving you confidence in your research.
Learn about the Conferma® ELISA development and manufacturing methods that provide strong sample detection and long-term assay and lot consistency, giving you confidence in your research.
Protocols
This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.
This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.
This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.
This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.
Related Content
Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.
Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service