Skip to Content
Merck
All Photos(2)

Key Documents

RAB0092

Sigma-Aldrich

Human hCG (intact) ELISA Kit

for serum, plasma, and cell culture supernatant

Synonym(s):

CG-beta, Chorionic gonadotrophin chain beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CGB(1082)

General description

The Human hCG (intact) (human Chorionic Gonadotropin) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human hCG (intact) in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human hCG

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human hCG (intact) ELISA Kit has been used in the detection of β-human chorionic gonadotropin in human plasma samples.

Biochem/physiol Actions

Human Chorionic Gonadotropin (hCG) produced by syncytiotrophoblast cells of the placenta, is crucial for the maintenance of pregnancy and corpus luteum functionality. Hyperglycosylated hCG (hCGh) not only promotes cell growth during implantation, but favors tumor growth and invasion in cancer. hCGh levels is indicative of malignancies like Down syndrome in pregnancies. hCG based vaccine has contraceptive functionality.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Advances in development of a contraceptive vaccine against human chorionic gonadotropin
Talwar GP, et al.
Expert opinion on biological therapy, 15(8), 1183-1190 (2015)
Analysis of the efficacy of resveratrol treatment in patients with scarred uterus
Ma H and Qiao Z
Experimental and Therapeutic Medicine, 15(6), 5410-5414 (2018)
Human choriogonadotrophin protein core and sugar branches heterogeneity: basic and clinical insights
De Medeiros, SF and Norman, RJ
Human Reproduction Update, 15(1), 69-95 (2008)
New discoveries on the biology and detection of human chorionic gonadotropin
Cole LA
Reproductive Biology and Endocrinology, 7(1), 8-8 (2009)
Analysis of the efficacy of resveratrol treatment in patients with scarred uterus
Ma H and Qiao Z
Experimental and Therapeutic Medicine, 15, 5410-5414 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service