RAB0002
Mouse ACE ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.12 ng/mL
standard curve range: 0.123-30 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Ace(11421)
General description
The Mouse ACE (Angiotensin-Converting Enzyme) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse ACE in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Mouse ACE
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Mouse ACE ELISA Kit has been used to quantify total serum concentration of angiotensin converting enzyme (ACE) in spontaneously hypertensive rats.
Biochem/physiol Actions
Angiotensin I converting enzyme (ACE) catalyzes the formation of angiotensin II from angiotensin I. ACE is a key aminopeptidase of the renin-angiotensin system. It is a membrane-bound enzyme which is expressed in atherosclerotic lesions.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Local corticosterone production and angiotensin-I converting enzyme shedding in a mouse model of intestinal inflammation
World Journal of Gastroenterology (2015)
Science translational medicine, 12(563) (2020-10-02)
Recent genome-wide association studies identified the angiotensin-converting enzyme gene (ACE) as an Alzheimer's disease (AD) risk locus. However, the pathogenic mechanism by which ACE causes AD is unknown. Using whole-genome sequencing, we identified rare ACE coding variants in AD families
Angiotensin-Converting Enzyme in Smooth Muscle Cells Promotes Atherosclerosis
Arteriosclerosis, Thrombosis, and Vascular Biology (2016)
Vaccines, 8(1) (2020-02-27)
Chagas cardiomyopathy is caused by Trypanosoma cruzi (Tc). We identified two candidate antigens (TcG2 and TcG4) that elicit antibodies and T cell responses in naturally infected diverse hosts. In this study, we cloned TcG2 and TcG4 in a nanovector and
A study on the effect of the Dendropanax mobifera extract on anti-hypertensive
Journal of the Korea Academia-Industrial cooperation Society, 17, 708-715 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service