Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NA.32
UNSPSC Code:
12352204
species reactivity
mouse, human, rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable, enzyme immunoassay: suitable
input
sample type cell lysate
sample type culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 2.62 pg/mL
standard curve range: 0.1-1000 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... AGT(183)
General description
The Angiotensin II Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Angiotensin II peptide based on the principle of Competitive Enzyme Immunoassay. EIA-ANGII-1 can theoretically detect all active angiotensins, including ANGI, ANGII, ANGIII and ANGIV. However, it does not detect inactive angiotensinogen.
Immunogen
Angiotensin II, synthetic peptide
Application
Angiotensin II EIA Kit has been used for quantitative estimation of serum angiotensin-II.
For research use only. Not for use in diagnostic procedures.
Angiotensin II EIA Kit has been used for analyzing circulating and serum levels of angiotensin II (Ang II) in mice and rats respectively.
Angiotensin II EIA Kit has been used for analyzing circulating and serum levels of angiotensin II (Ang II) in mice and rats respectively.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Study on in vitro model shows that mechanical stretch induced Ang II release plays a vital role in mediating stretch-induced hypertrophic response. Ang II is implicated in maintaining blood pressure and has an important role in hypertension and in the pathophysiology of cardiovascular disorders. Ang II also has an essential role in growth induction, cell migration and mitosis of vascular smooth muscle cells. It enhances the production of collagen type I and III in fibroblasts, that results in thickening of the vascular wall and myocardium, and fibrosis. Variation in Ang II gene increases the risk of myocardial infarction, hypertension and left ventricular hypertrophy. Ang II expression is associated with heart failure, cardiac remodeling, diabetes, and the proliferative and inflammatory responses to arterial injury. The pharmacological agents such as angiotensin II-converting enzyme inhibitors or angiotensin II receptor antagonists can be used for treating Ang II-induced hypertension and congestive heart failure.
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Genetic abrogation of adenosine A3 receptor prevents uninephrectomy and high salt-induced hypertension
Yang T, et al.
Journal of the American Heart Association, 5(7), e003868-e003868 (2016)
Autocrine release of angiotensin II mediates stretch-induced hypertrophy of cardiac myocytes in vitro.
Sadoshima J
Cell, 75(5), 977-984 (1993)
Daniela Volonte et al.
The Journal of biological chemistry, 296, 100242-100242 (2021-01-01)
Oncogenic K-Ras (K-RasG12V) promotes senescence in normal cells but fuels transformation of cancer cells after the senescence barrier is bypassed. The mechanisms regulating this pleiotropic function of K-Ras remain to be fully established and bear high pathological significance. We find
Genetic Abrogation of Adenosine A3 Receptor Prevents Uninephrectomy and High Salt-Induced Hypertension.
Yang T
Journal of the American Heart Association, 5(7) (2016)
Gagandeep Kaur et al.
Frontiers in physiology, 12, 653045-653045 (2021-06-15)
Background: Tobacco smoking is known to be involved in the pathogenesis of several cardiopulmonary diseases. Additionally, smokers are highly susceptible to infectious agents due to weakened immunity. However, the progression of lung injury based on SARS-CoV-2-mediated COVID-19 pathogenesis amongst smokers
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| RAB0010-1KT | 04061834959021 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service