Skip to Content
Merck
All Photos(1)

Documents

R7509

Sigma-Aldrich

RPMI-1640 Medium

With sodium bicarbonate, without ʟ-glutamine and phenol red, liquid, sterile-filtered,suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: no
L-glutamine: no
HEPES: no
sodium pyruvate: no
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.
RPMI-1640 Medium is suitable for culturing human normal and neoplastic leukocytes. 

Application

RPMI-1640 Medium has been used:
  • to culture Mantle cell lymphoma cells
  • to culture mouse podocytes
  • in the extraction of whole bone marrow from rat Femur and tibia bones

Other Notes

Phenol red has been shown to interfere with the growth of some cells at clonal densities.
Use this medium when working with stem cells or when growing cells at low densities.
Also recommended for in vitro diagnostics use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingvild S M Gabrielsen et al.
PloS one, 14(7), e0218858-e0218858 (2019-07-02)
Antigen presenting cells (APCs) in the thymus play an essential role in the establishment of central tolerance, i.e. the generation of a repertoire of functional and self-tolerant T cells to prevent autoimmunity. In this study, we have compared the transcriptomes
Ida Marie Rundgren et al.
Journal of immunological methods, 461, 53-62 (2018-06-16)
Monocytes are important for innate immunity and include the classical (CD14brightCD16negative), intermediate (CD14brightCD16dim) and non-classical (CD14dimCD16bright) monocyte subsets. The quantification of these functionally different subsets in peripheral blood may become useful for diagnosis and follow-up in human diseases. The aim
Alyson A Fiorillo et al.
Frontiers in immunology, 11, 151-151 (2020-03-11)
MicroRNAs (miRNAs) are small non-coding RNA molecules that regulate important intracellular biological processes. In myasthenia gravis (MG), a disease-specific pattern of elevated circulating miRNAs has been found, and proposed as potential biomarkers. These elevated miRNAs include miR-150-5p, miR-21-5p, and miR-30e-5p
Timurs Maculins et al.
Scientific reports, 6, 18940-18940 (2016-01-09)
Ubiquitin signalling regulates most aspects of cellular life, thus deregulation of ubiquitylation has been linked with a number of diseases. E3 ubiquitin ligases provide substrate selectivity in ubiquitylation cascades and are therefore considered to be attractive targets for developing therapeutic
Deepak Kumar et al.
Aging, 14(5), 2081-2100 (2022-03-02)
Alterations in RNA splicing are associated with different malignancies, including leukemia, lymphoma, and solid tumors. The RNA splicing modulators such as FD-895 and pladienolide B have been investigated in different malignancies to target/modulate spliceosome for therapeutic purpose. Different cell lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service