Skip to Content
Merck
All Photos(2)

Key Documents

R7257

Sigma-Aldrich

Ricinoleic acid

≥95%

Synonym(s):

Ricinelaidic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH2CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
298.46
Beilstein:
1727813
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

natural (organic)

Assay

≥95%

form

liquid

density

0.940 g/mL at 20 °C (lit.)

functional group

carboxylic acid
hydroxyl

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h9,12,17,19H,2-8,10-11,13-16H2,1H3,(H,20,21)/b12-9-/t17-/m1/s1

InChI key

WBHHMMIMDMUBKC-QJWNTBNXSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Multiplexed PLGA scaffolds with nitric oxide-releasing zinc oxide and melatonin-modulated extracellular vesicles for severe chronic kidney disease.: This innovative study utilizes ricinoleic acid in the fabrication of multiplexed PLGA scaffolds, demonstrating its effectiveness in enhancing the therapeutic efficacy of treatments for chronic kidney disease. The research underscores the potential of ricinoleic acid in medical applications, particularly in regenerative medicine and drug delivery systems (Rhim WK et al., 2024).

  • Optimizing the enzymatic production of biolubricants by the Taguchi method: Esterification of the free fatty acids from castor oil with 2-ethyl-1-hexanol catalyzed by Eversa Transform 2.0.: Demonstrates the industrial application of ricinoleic acid in producing biolubricants, emphasizing its economic and environmental benefits. The study provides insights into the sustainable production processes and industrial applications of ricinoleic acid (Monteiro RRC et al., 2024).

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

435.2 °F - closed cup

Flash Point(C)

224 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuanyi Liang et al.
Food research international (Ottawa, Ont.), 134, 109237-109237 (2020-06-11)
This study investigated the relationships between the structures of hydroxy unsaturated fatty acids (HUFA) and their antifungal activities. Structurally diverse HUFA, including four monohydroxy-18:1 isomers, two monohydroxy 18:2 isomers and two monohydroxy 18:2 isomers were extracted from seeds of plants
Petra Kocbek et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 50(1), 130-138 (2013-04-23)
Superparamagnetic iron oxide nanoparticles (SPIONs) show a great promise for a wide specter of bioapplications, due to their characteristic magnetic properties exhibited only in the presence of magnetic field. Their advantages in the fields of magnetic drug targeting and imaging
Megan L Robertson et al.
ACS applied materials & interfaces, 3(9), 3402-3410 (2011-08-10)
Poly(l-lactide) (PLLA) is a renewable resource polymer derived from plant sugars with several commercial applications. Broader implementation of the material is limited due to its inherent brittleness. We show that the addition of 5 wt % castor oil to PLLA
Ioannis Mavraganis et al.
Applied and environmental microbiology, 76(4), 1135-1142 (2009-12-22)
Claviceps purpurea, the fungal pathogen that causes the cereal disease ergot, produces glycerides that contain high levels of ricinoleic acid [(R)-12-hydroxyoctadec-cis-9-enoic acid] in its sclerotia. Recently, a fatty acid hydroxylase (C. purpurea FAH [CpFAH]) involved in the biosynthesis of ricinoleic
André Arnosti et al.
Experimental parasitology, 127(2), 575-580 (2010-11-13)
This study examines the effects of ricinoleic acid esters from Ricinus communis castor oil on the vitellogenesis of Rhipicephalus sanguineus ticks attached to hosts that were fed with commercial rabbit food containing these esters. The oocytes of ticks from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service