Skip to Content
Merck
All Photos(1)

Key Documents

R3653

Sigma-Aldrich

Anti-Radixin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DFNB24

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

mouse, rat, human

technique(s)

immunoprecipitation (IP): 10-20 μg using RIPA extract (250 μg) of rat liver
indirect immunofluorescence: 10-20 μg/mL using cultured human HeLa cells
western blot: 1-2 μg/mL using whole extract of cultured HeLa cells
western blot: 1-2 μg/mL using whole extract of cultured mouse NIH3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RDX(5962)
mouse ... Rdx(19684)
rat ... Rdx(315655)

General description

RDX (radixin) belongs to the ERM (ezrin-radixin-moesin) family. It is present in several normal cells, neoplastic cells, epithelial and lymphoid cells. It is located on chromosome 11. In mouse, RDX is found in the hair cell stereocilia of inner ear.

Immunogen

synthetic peptide corresponding to amino acid residues 400-409 of human radixin with an N-terminal added tripeptide, conjugated to KLH. The corresponding sequence is identical in mouse, rat, and pig and differs by two amino acids in chicken.

Application

Anti-Radixin antibody produced in rabbit has been used in
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence staining of cultured cells

Biochem/physiol Actions

In human, knockdown of radixin (RDX) represses the development of glioblastoma cell in vitro and in vivo. It participates in cell motility, invasion and tumor progression. It acts as a membrane-cytoskeletal linker in cell surface structures that are abundant with actin. It is necessary for the arrangement of cortical cytoskeleton. RDX mutation can lead to DFNB24 hearing loss in human.
Radixin is the dominant ezrin-radixin-moesin (ERM) protein in liver bile canaliculi where it is considered to have a critical role in bile conjugated-bilirubin secretion by influencing the cellular localization of the multidrug resistance protein 2 (MRP2). Resting normal human blood cells lack detectable radixin. Radixin is found in classical NK cells. Radixin has closed and open forms corresponding to the inactive and active forms as cross-linkers between actin filaments. It may be phosphorylated and it plays an important role in the activation of the Rho family members by recruiting their positive and negative regulators.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radixin knockdown by RNA interference suppresses human glioblastoma cell growth in vitro and in vivo
Qin JJ, et al.
Asian Pacific Journal of Cancer Prevention, 15(22), 9805-9812 (2014)
Radixin regulates cell migration and cell-cell adhesion through Rac1.
Valderrama F, et al.
Journal of Cell Science, 125(14), 3310-3319 (2012)
Differential expression and distribution of ezrin, radixin and moesin in human natural killer cells.
Ramoni C, et al.
European Journal of Immunology, 32(11), 3059-3065 (2002)
Kuu Ikäheimo et al.
Life science alliance, 5(2) (2021-11-25)
Failure in the structural maintenance of the hair cell stereocilia bundle and ribbon synapse causes hearing loss. Here, we have studied how ER stress elicits hair cell pathology, using mouse models with inactivation of Manf (mesencephalic astrocyte-derived neurotrophic factor), encoding
Structural conversion between open and closed forms of radixin: low-angle shadowing electron microscopy.
Ishikawa H, et al.
Journal of molecular biology, 310(5), 973-978 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service