Skip to Content
Merck
All Photos(1)

Documents

Q3638

Sigma-Aldrich

Quetiapine hemifumarate salt

≥98% (HPLC)

Synonym(s):

2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol hemifumarate, Quetiapine fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N3O2S · 0.5C4H4O4
CAS Number:
Molecular Weight:
441.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24.OCCOCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68

InChI

1S/2C21H25N3O2S.C4H4O4/c2*25-14-16-26-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)27-20-8-4-2-6-18(20)22-21;5-3(6)1-2-4(7)8/h2*1-8,25H,9-16H2;1-2H,(H,5,6)(H,7,8)/b;;2-1+

InChI key

ZTHJULTYCAQOIJ-WXXKFALUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Quetiapine is a derivative of dibenzothiazepine compound and is also referred to as seroquel and clemastine.

Application

Quetiapine hemifumarate salt has been used as an antagonist for β-arrestin 2 mutant T205M recruitment.

Biochem/physiol Actions

Quetiapine hemifumarate is an atypical antipsychotic, a combined serotonin (5HT2) and dopamine (D2) receptor antagonist.
Quetiapine promotes oligodendrogenesis in vitro. It is used in treating depressive disorders.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of rat oligodendrocyte progenitor cultures and quantification of oligodendrogenesis using dual-infrared fluorescence scanning
Schott JT, et al.
Journal of Visualized Experiments, 108, e53764-e53764 (2016)
Meng He et al.
Frontiers in aging neuroscience, 14, 847561-847561 (2022-05-27)
Emerging data indicate that antipsychotic treatment causes brain volume loss and astrocyte death, but the mechanisms remain elusive. Pyroptosis, inflammatory cell death characterized by the formation of inflammatory bodies, increased expression of nod-like receptor proteins (NLRPs) such as NLRP3, and
Marc F Goldstein et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 97(6), 717-730 (2007-01-05)
To review and compare previously reported cases of selective IgM immunodeficiency (SIgMID) with the largest adult cohort obtained from a retrospective analysis of an allergy and immunology practice. Publications were selected from the English-only PubMed database (1966-2005) using the following
Ella J Daly et al.
Neuropsychiatric disease and treatment, 3(6), 855-867 (2007-12-01)
Atypical antipsychotics are increasingly used in the treatment of a broad spectrum of psychiatric disorders. There is evidence that in addition to treating the positive and negative symptoms of schizophrenia, as well as mania in bipolar disorder, these agents may
Jason T Schott et al.
Journal of visualized experiments : JoVE, (108), 53764-53764 (2016-03-12)
Efficient oligodendrogenesis is the therapeutic goal of a number of areas of research including spinal cord injury, neonatal hypoxia, and demyelinating diseases such as multiple sclerosis and transverse myelitis. Myelination is required to not only facilitate rapid impulse propagation within

Articles

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service