Skip to Content
Merck
All Photos(1)

Documents

PZ038

Sigma-Aldrich

PF-06928215

≥98% (HPLC)

Synonym(s):

rel-(1R,2S)2(7-Oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3 carboxamido) cyclohexane-1-carboxylic acid, rel-(1R,2S)2(7-Oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3-carboxamido)cyclohexanecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N4O4
CAS Number:
Molecular Weight:
380.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

Application

PF-06928215 has been used to study its inhibitory effects on cyclic GMP-AMP synthase (cGAS).

Biochem/physiol Actions

PF-06928215 is a high affinity (KD = 200 nM) active site-targeting, substrate-competitive cyclic GMP-AMP synthase (cGAS) inhibitor (IC50 = 4.9 μM; in the presence of 1 mM ATP, 0.3 mM GTP, 100 nM 45-bp interferon-stimulatory dsDNA (ISD)). PF-06928215 displays no inhibitory potency against dsDNA-induced IFN-β expression in cellular cGAS assays, most likely due to limited cell-permeability and/or high levels of cellular ATP and GTP in the reporter cells employed.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of PF-06928215 as a high affinity inhibitor of cGAS enabled by a novel fluorescence polarization assay
Hall J, et al.
PLoS ONE, 12(9), e0184843-e0184843 (2017)
Subhadip Choudhuri et al.
PLoS pathogens, 16(4), e1008474-e1008474 (2020-04-22)
Trypanosoma cruzi (T. cruzi) is the etiological agent of Chagas cardiomyopathy. In the present study, we investigated the role of extracellular vesicles (Ev) in shaping the macrophage (Mφ) response in progressive Chagas disease (CD). We purified T. cruzi Ev (TcEv)
Structure of the human cGAS-DNA complex reveals enhanced control of immune surveillance
Zhou W, et al.
Cell, 174(2), 300-311 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service