Skip to Content
Merck
All Photos(1)

Key Documents

P6617

Sigma-Aldrich

Pristanic acid

≥97% (GC), ethanol solution

Synonym(s):

2,6,10,14-Tetramethylpentadecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38O2
CAS Number:
Molecular Weight:
298.50
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Product Name

Pristanic acid solution, mixture of isomers, ethanol solution, ≥97% (GC)

Quality Level

Assay

≥97% (GC)

form

ethanol solution

storage temp.

−20°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC(C)C(O)=O

InChI

1S/C19H38O2/c1-15(2)9-6-10-16(3)11-7-12-17(4)13-8-14-18(5)19(20)21/h15-18H,6-14H2,1-5H3,(H,20,21)

InChI key

PAHGJZDQXIOYTH-UHFFFAOYSA-N

Biochem/physiol Actions

Oxidation product of fatty acids in the diet, particularly phytanic acid. Phytanic acid is oxidized by alpha oxidation yielding pristanic acid, which is subsequently degraded by peroxisomal beta oxidation. Several inborn errors of metabolism with one or more deficiencies in the phytanic acid and pristanic acid breakdown have been described.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ferdinandusse et al.
American journal of human genetics, 78(6), 1046-1052 (2006-05-11)
In this report, we describe the first known patient with a deficiency of sterol carrier protein X (SCPx), a peroxisomal enzyme with thiolase activity, which is required for the breakdown of branched-chain fatty acids. The patient presented with torticollis and
Antonella Peduto et al.
Molecular genetics and metabolism, 82(3), 224-230 (2004-07-06)
Peroxisomal disorders include a complex spectrum of diseases, characterized by a high heterogeneity from both the clinical and the biochemical points of view. Specific assays are required for the study of peroxisome metabolism. Among these, pipecolic acid evaluation is considered
J Gootjes et al.
Neurology, 59(11), 1746-1749 (2002-12-11)
To identify prognostic markers reflecting the extent of peroxisome dysfunction in primary skin fibroblasts from patients with peroxisome biogenesis disorders (PBD). PBD are a genetically heterogeneous group of disorders due to defects in at least 11 distinct genes. Zellweger syndrome
Biochemical markers predicting survival in peroxisome biogenesis disorders.
Jeannette Gootjes et al.
Advances in experimental medicine and biology, 544, 67-68 (2004-01-10)
Phytanic and pristanic acid are naturally occuring ligands.
Anna W M Zomer et al.
Advances in experimental medicine and biology, 544, 247-254 (2004-01-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service