Skip to Content
Merck
All Photos(1)

Documents

PZ0007

Sigma-Aldrich

Azithromycin dihydrate

≥98% (HPLC), powder, 50S ribosomal subunit formation and elongation inhibitor

Synonym(s):

1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10- trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylohexopyranosyl]oxy]-, dihydrate, [2R-(2R, 3S, 4R, 5R, 8R, 10R, 11R, 12S, 13S, 14R)]-, N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A, CP-62993

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H72N2O12 · 2H2O
CAS Number:
Molecular Weight:
785.02
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Azithromycin dihydrate, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

O.O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C38H72N2O12.2H2O/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28;;/h20-33,35,41-43,45-46H,15-19H2,1-14H3;2*1H2/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-;;/m1../s1

InChI key

SRMPHJKQVUDLQE-KUJJYQHYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Azithromycin dihydrate has been used:
  • in blocking buffer (5% bovine serum albumin) for indirect immunofluorescence
  • in treating the U87 cells for cell-viability assay and flow cytometry analysis to study the Zika virus infection in glial cells
  • in antibiotic co-treatment experiments, to compare its antibacterial activity with PYRRO-C3D against Streptococcus pneumoniae biofilms

Biochem/physiol Actions

Azithromycin dihydrate is a macrolide antibiotic, azalide subclass. It binds to the 50S subunit of the 70S bacterial ribosomes and inhibits RNA-dependent protein synthesis in bacterial cells. Azithromycin also has anti-immunomodulatory/anti-inflammatory properties, which make it useful in treating cystic fibrosis.
Azithromycin is a non-β-lactam antibody, effective against infections associated with respiratory tract, skin, tissues and genital chlamydia. It also acts against Plasmodium falciparum and P. vivax, which are multidrug resistance species that transmit malaria. Azithromycin targets the ribosome and prevents protein synthesis. It is soluble in lipids and metabolizes in liver by undergoing demethylation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zika virus cell tropism in the developing human brain and inhibition by azithromycin
Retallack H, et al.
Proceedings of the National Academy of Sciences of the USA, 113(50), 14408-14413 (2016)
Human Sertoli cells support high levels of Zika virus replication and persistence
Kumar A, et al.
Scientific Reports, 8(1), 5477-5477 (2018)
Travelers' Malaria, 131-131 (2008)
Alexander J Currie et al.
Frontiers in cellular and infection microbiology, 10, 372-372 (2020-08-15)
Excessive inflammation by phagocytes during Aspergillus fumigatus infection is thought to promote lung function decline in CF patients. CFTR modulators have been shown to reduce A. fumigatus colonization in vivo, however, their antifungal and anti-inflammatory mechanisms are unclear. Other treatments
Paul N Newton et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 68(5), 738-747 (2018-07-19)
Murine typhus, or infection with Rickettsia typhi, is a global but neglected disease without randomized clinical trials to guide antibiotic therapy. A prospective, open, randomized trial was conducted in nonpregnant, consenting inpatient adults with rapid diagnostic test evidence of uncomplicated

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service