Skip to Content
Merck
All Photos(1)

Key Documents

P9273

Sigma-Aldrich

Pro-Phe-Arg-7-amido-4-methylcoumarin

≥95%

Synonym(s):

PFR-Nmec

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37N7O5 · C2H4O2
CAS Number:
Molecular Weight:
635.71
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95%

form

powder

composition

Peptide content, ≥65%

solubility

methanol: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

NC(NCCC[C@@H](C(NC1=CC=C(C(C)=CC(O2)=O)C2=C1)=O)NC([C@H](CC3=CC=CC=C3)NC(C4NCCC4)=O)=O)=N.CC([O-])=O

InChI

1S/C30H37N7O5.ClH/c1-18-15-26(38)42-25-17-20(11-12-21(18)25)35-28(40)23(10-6-14-34-30(31)32)36-29(41)24(16-19-7-3-2-4-8-19)37-27(39)22-9-5-13-33-22;/h2-4,7-8,11-12,15,17,22-24,33H,5-6,9-10,13-14,16H2,1H3,(H,35,40)(H,36,41)(H,37,39)(H4,31,32,34);1H

InChI key

QYFPUTDFLKFOBA-UHFFFAOYSA-N

Application

Pro-Phe-Arg-7-amido-4-methylcoumarin has been used as fluorogenic substrate:
  • in kinetic experiments to measure the enzymatic activity of factor XII
  • to measure the activity of generated plasma kallikrein using Spectramax plate reader
  • to determine the cathepsin A (CatA) inhibitor selectivity against various proteases using Tecan Safire 2 plate reader

Substrates

A fluorogenic substrate for pancreatic and urinary kallikreins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Paul et al.
Journal of immunology (Baltimore, Md. : 1950), 159(3), 1530-1536 (1997-08-01)
Polyreactive and thyroglobulin (Tg)-directed proteolytic activities present in the serum IgG of healthy controls and patients with autoimmune disease were studied by electrophoretic separation of 125I-labeled Tg reaction products and spectrofluorometric measurement of Pro-Phe-Arg-methylcoumarinamide cleavage at the Arg-methylcoumarinamide bond. A
K Uchida et al.
Biochimica et biophysica acta, 614(2), 501-510 (1980-08-07)
Kallikrein (EC 3.4.21.8) was purified from rat stomach by column chromatography on p-aminobenzamidine-Sepharose, DEAE-Sephadex A-50 and Sephadex G-150 and by isoelectric focusing, measuring its activities to hydrolyse L-prolyl-L-phenylalanyl-L-arginine-4-methyl-coumaryl-7-amide and to release kinin from heat-treated rat plasma. the purified stomach kallikrein
Sven Ruf et al.
Journal of medicinal chemistry, 55(17), 7636-7649 (2012-08-07)
Cathepsin A (CatA) is a serine carboxypeptidase distributed between lysosomes, cell membrane, and extracellular space. Several peptide hormones including bradykinin and angiotensin I have been described as substrates. Therefore, the inhibition of CatA has the potential for beneficial effects in
Y Kitamura et al.
Biochimica et biophysica acta, 1387(1-2), 153-164 (1998-09-28)
The culture medium in which prehatching embryos of the frog, Rana pirica, were cultured (hatching medium) solubilized the vitelline coat (VC) of unfertilized eggs and contained molecules reactive to antibodies (anti-UVS.2) against the Xenopus hatching enzyme (HE). The hydrolyzing activity
L Li et al.
Clinical and experimental immunology, 120(2), 261-266 (2000-05-03)
Chemical catalysis, an effector mechanism utilized by fully assembled antibodies, can also be mediated by the isolated antibody subunits. Because trace amounts of free light chains (L chains) are present in IgG preparations, a detailed study was undertaken to identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service