Skip to Content
Merck
All Photos(3)

Key Documents

P8759

Sigma-Aldrich

Pyrogallol Red

Powder

Synonym(s):

Pyrogallolsulfonephthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12O8S
CAS Number:
Molecular Weight:
400.36
Beilstein:
61394
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Pyrogallol Red, Suitable for use as a complexometric indicator

Quality Level

form

powder

technique(s)

titration: suitable

color

green to very dark green, and brown

mp

≥300 °C (lit.)

solubility

1 M NH4OH: 10 mg/mL, clear

εmax

≥450 at 475 nm in methanol

suitability

suitable for complexometric indicator

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc2c(Oc3c(O)c(O)ccc3C24OS(=O)(=O)c5ccccc45)c1O

InChI

1S/C19H12O8S/c20-12-7-5-10-17(15(12)22)26-18-11(6-8-13(21)16(18)23)19(10)9-3-1-2-4-14(9)28(24,25)27-19/h1-8,20-23H

InChI key

KUQNCHZOCSYKOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrogallol red (PGR) is an ortho-diphenolic dye. It has high reactivity towards reactive oxygen species (ROS) and reactive nitrogen species (RNS) that include peroxynitrite, radical, peroxyl radicals, hypochlorite & superoxide anion.

Application

Pyrogallol Red is suitable to prepare Fujita and Watanabe dye reagent used in Pyrogallol Red-Molybdenum (IV) complex (PR) method for the determination and quantification of urinary proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tuula K Outinen et al.
Infectious diseases (London, England), 49(11-12), 840-846 (2017-08-02)
Puumala hantavirus (PUUV) causes haemorrhagic fever with renal syndrome characterized by thrombocytopenia, capillary leakage and acute kidney injury (AKI) with proteinuria and haematuria. Although the typical histologic lesion is acute tubulointerstitial nephritis, the amount of glomerular proteinuria predicts the severity
Nicolas Herman et al.
Journal of veterinary internal medicine, 33(2), 999-1008 (2019-02-16)
There are no reference intervals for urinalysis in cattle. Characterize the urine of healthy cows, establish urine protein-to-creatinine ratio (UPC) reference intervals, and test possible differences among dairy and beef cattle, age groups, or stage of lactation. Seventy-seven dairy and
K Janšáková et al.
Physiological research, 68(3), 467-479 (2019-03-25)
Thermally processed food contains advanced glycation end products (AGEs) including N(epsilon)-(carboxymethyl)lysine (CML). Higher AGEs or circulating CML were shown to be associated with pregnancy complications such as preeclampsia and gestational diabetes. It is unclear whether this association is causal. The
K de Leeuw et al.
Journal of critical care, 36, 200-206 (2016-11-05)
In patients with severe burns, resuscitation with large volumes of fluid is needed, partly because of an increase in capillary leakage. Corticosteroids might be beneficial by diminishing capillary leakage. This study aimed to assess in severely burned nonseptic patients whether
Yo Omata et al.
Journal of agricultural and food chemistry, 56(9), 3386-3390 (2008-04-09)
The radical-scavenging antioxidants play an important role against oxidative stress in the defense system in vivo. The beneficial effects of antioxidants contained in foods and beverages have been well-accepted, and their antioxidant capacity has been assessed by various methods. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service