Skip to Content
Merck
All Photos(2)

Key Documents

P8165

Sigma-Aldrich

Phosphate Buffer pH 6.6 at 25 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.47
Pricing and availability is not currently available.

form

powder

color

white

pH

6.60-6.80

application(s)

hematology
histology

storage temp.

room temp

General description

Phosphate buffer′s buffering capacity is pH sensitive and is preferred to be used at physiological pH (7.2-7.4).

Application

For use with Wright Stain for Geometric Data HEMASTAIN®.

Phosphate buffer serves as an important fixative for light and electron microscopy. It is the most commonly used component of irrigation solution during surgeries, to preserve living tissues. It is also used in immunochemical staining procedures.

Reconstitution

Each vial will prepare 3.8L of buffer, 34mM.

Legal Information

HEMASTAIN is a registered trademark of Smithkline Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Biological Electron Microscopy: Theory, Techniques, and Troubleshooting (2011)

Questions

  1. What is the weight of the product? Is it 35.3g?

    1 answer
    1. The weight of the individual vials is not recorded. Although the weights for random bottles are determined, the weights for each vial are not recorded. It is expected that each vial should contain approximately 17.5 grams, and anything between 17.5 to 18 grams would be considered acceptable.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service