Skip to Content
Merck
All Photos(1)

Documents

H1792

Sigma-Aldrich

Hyperforin (dicyclohexylammonium) salt

Synonym(s):

Hyp-DCHA, Hyperforin-DCHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H52O4·C12H23N
CAS Number:
Molecular Weight:
718.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)C(=O)[C@@]34C(O)=C(C\C=C(\C)C)C(=O)[C@@](C\C=C(\C)C)(C[C@H](C\C=C(/C)C)[C@@]3(C)CC\C=C(\C)C)C4=O

InChI

1S/C35H52O4.C12H23N/c1-22(2)13-12-19-33(11)27(16-14-23(3)4)21-34(20-18-25(7)8)30(37)28(17-15-24(5)6)31(38)35(33,32(34)39)29(36)26(9)10;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h13-15,18,26-27,38H,12,16-17,19-21H2,1-11H3;11-13H,1-10H2/t27-,33+,34+,35-;/m0./s1

InChI key

ZUYCAROZOLUHJY-DNSWOBEMSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort. It acts as a reuptake inhibitor of monoamines, including serotonin, norepinephrine, dopamine, and of GABA and glutamate by activating the transient receptor potential ion channel TRPC6. It also is an activator of the pregnane X receptor (PXR), which serves as a key regulator of CYP3A4 transcription, a member of the cytochrome (CYP) P450 enzyme system. Recent studies have found it to have neuroprotective effects against Alzheimer′s disease (AD) neuropathology, including the ability to disassemble amyloid-β aggregates in vitro and improve spatial memory in vivo.
Stable salt of hyperforin. Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort. It acts as a reuptake inhibitor of monoamines, including serotonin, norepinephrine, dopamine, and of GABA and glutamate by activating the transient receptor potential ion channel TRPC6. Activation of TRPC6 induces the entry of sodium and calcium into the cell which causes inhibition of monoamine reuptake. It also is an activator of the pregnane X receptor (PXR), which serves as a key regulator of CYP3A4 transcription, a member of the cytochrome (CYP) P450 enzyme system. Recent studies have found other neurological effects, effects on inflammation, as well as antibacterial, antitumoral and antiangiogenic effects.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong Hee Jeon et al.
Lung cancer (Amsterdam, Netherlands), 69(2), 165-171 (2009-12-01)
Lipid raft, a specialized membrane structure enriched with cholesterol and glycosphingolipid, contains molecules that convey environmental stimuli to the intracellular systems. Authors investigated the effects of raft cholesterol depletion on non-small cell lung cancer (NSCLC) cell migration. Incubation of NSCLC
Ying Zheng et al.
Cancer research, 69(3), 896-904 (2009-01-30)
Cyclooxygenase-2-derived prostaglandin E(2) (PGE(2)) stimulates tumor cell growth and progression. However, the mechanisms by which PGE(2) increases tumor growth remain incompletely understood. In studies performed in non-small cell lung carcinoma (NSCLC) cells, we found that PGE(2) stimulates the expression of
Noriaki Sunaga et al.
Molecular cancer therapeutics, 10(2), 336-346 (2011-02-11)
Oncogenic KRAS is found in more than 25% of lung adenocarcinomas, the major histologic subtype of non-small cell lung cancer (NSCLC), and is an important target for drug development. To this end, we generated four NSCLC lines with stable knockdown
Jacqueline C Shultz et al.
Cancer research, 70(22), 9185-9196 (2010-11-04)
Increasing evidence points to the functional importance of alternative splice variations in cancer pathophysiology. Two splice variants are derived from the CASP9 gene via the inclusion (Casp9a) or exclusion (Casp9b) of a four-exon cassette. Here we show that alternative splicing
Jochen Kühnl et al.
Toxicology, 448, 152637-152637 (2020-11-22)
Microphysiological systems (MPS) aim to mimic the dynamic microenvironment and the interaction between tissues. While MPS exist for investigating pharmaceuticals, the applicability of MPS for cosmetics ingredients is yet to be evaluated. The HUMIMIC Chip2 ("Chip2″), is the first multi-organ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service