Skip to Content
Merck
All Photos(4)

Key Documents

P6503

Sigma-Aldrich

Pilocarpine hydrochloride

≥99% (titration), powder, muscarinic acetylcholine receptor agonist

Synonym(s):

(3S,4R)-4,5-Dihydro-3-ethyl-4-(1-methyl-1H-imidazol-5-ylmethyl)-2(3H)-furanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · HCl
CAS Number:
Molecular Weight:
244.72
Beilstein:
4034491
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Pilocarpine hydrochloride, ≥99% (titration), powder

Quality Level

Assay

≥99% (titration)

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

202-205 °C (lit.)

solubility

H2O: 100 mg/mL

application(s)

forensics and toxicology
veterinary

relevant disease(s)

epilepsy

SMILES string

Cl.CC[C@H]1[C@H](COC1=O)Cc2cncn2C

InChI

1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1

InChI key

RNAICSBVACLLGM-GNAZCLTHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pilocarpine hydrochloride has been used to stimulate salivary secretions in mice. It has also been used to induce status epilepticus(SE) in rat.

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.
Pilocarpine is alkaloid isolated from the shrub Pilocarpus species. It comprises imidazole ring and γ- lactone rings in its structure. It has cholinergic potential and it stimulates the parasympathetic system. Pilocarpine is also effective for treating glaucoma. It stimulates ciliary muscle contraction by binding to muscarinic M3 receptors. It is effective in treating corneal dryness in Sjogren′s syndrome. It is a secretagogue, which improves salivary secretions in Xerostomia or a dry mouth condition in Sjogren′s syndrome patients and in radiotherapy patients with head and neck carcinoma.

Features and Benefits

This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic, light sensitive

Other Notes

Shelf life of the powder is at least five years.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Double-blind randomized, placebo-controlled study of pilocarpine to salvage salivary gland function during radiotherapy of patients with head and neck cancer
Gornitsky M, et al.
Oral Surgery, Oral Medicine, Oral Pathology, Oral Radiology, and Endodontics, 98(1), 45-52 (2004)
Biomarker profiles in serum and saliva of experimental Sjogren's syndrome: associations with specific autoimmune manifestations
Delaleu N, et al.
Arthritis Research & Therapy, 10(1), R22-R22 (2008)
Oral pilocarpine for the treatment of ocular symptoms in patients with Sjogren?s syndrome: a randomised 12 week controlled study
Tsifetaki N, et al.
Annals of the Rheumatic Diseases, 62(12), 1204-1207 (2003)
Longitudinal microPET imaging of brain glucose metabolism in rat lithium-pilocarpine model of epilepsy
Goffin K, et al.
Experimental neurology, 217(1), 205-209 (2009)
In vivo mapping of temporospatial changes in glucose utilization in rat brain during epileptogenesis: an 18F-fluorodeoxyglucose-small animal positron emission tomography study
Guo Y, et al.
Neuroscience, 162(4), 972-979 (2009)

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service