Skip to Content
Merck
All Photos(1)

Key Documents

H0261

Sigma-Aldrich

DL-β-Hydroxybutyryl coenzyme A lithium salt

≥90%

Synonym(s):

DL-3- Hydroxybutyryl coenzyme A lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H42N7O18P3S
CAS Number:
Molecular Weight:
853.62
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (Organic)

Assay

≥90%

form

powder

solubility

H2O: soluble-50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Li].CC(O)CC(=O)SCCNC(=O)CCNC(=O)C(O)C(C)(C)COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)n2cnc3c(N)ncnc23

InChI

1S/C25H42N7O18P3S.Li.H/c1-13(33)8-16(35)54-7-6-27-15(34)4-5-28-23(38)20(37)25(2,3)10-47-53(44,45)50-52(42,43)46-9-14-19(49-51(39,40)41)18(36)24(48-14)32-12-31-17-21(26)29-11-30-22(17)32;;/h11-14,18-20,24,33,36-37H,4-10H2,1-3H3,(H,27,34)(H,28,38)(H,42,43)(H,44,45)(H2,26,29,30)(H2,39,40,41);;

InChI key

HNPHARRBSFXZAL-UHFFFAOYSA-N

Application

DL-β-Hydroxybutyryl coenzyme A lithium salt has been used:

  • as a substrate in the HSD17B4 protein enzymatic assay
  • as a substrate to determine the specificity and kinetics of Hc-DHS-28 and its mutation proteins
  • as a component of the reaction buffer in S-adenosyl-L-homocysteine hydrolase (AHCY) in vitro acylation

Biochem/physiol Actions

3-Hydroxybutyryl coenzyme A (HBCoA) is converted to bacterial polyhydroxyalkanoates (PHB) by polyhydroxybutyrate (PHB) synthases. 3-Hydroxybutyryl coenzyme A is a substrate used to measure the specificity and kinetics of β-hydroxyacyl CoA dehydrogenase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kesaven Bhubalan et al.
Applied and environmental microbiology, 77(9), 2926-2933 (2011-03-15)
The synthesis of bacterial polyhydroxyalkanoates (PHA) is very much dependent on the expression and activity of a key enzyme, PHA synthase (PhaC). Many efforts are being pursued to enhance the activity and broaden the substrate specificity of PhaC. Here, we
Ping Li et al.
Biochemistry, 48(39), 9202-9211 (2009-08-29)
Polyhydroxybutyrate (PHB) synthases catalyze the conversion of 3-hydroxybutyryl coenzyme A (HBCoA) to PHB with a molecular mass of 1.5 MDa. The class III synthase from Allochromatium vinosum is a tetramer of PhaEPhaC (each 40 kDa). The polymerization involves covalent catalysis
Yi Yang et al.
International journal for parasitology, 50(12), 945-957 (2020-08-29)
Haemonchus contortus could enter the diapause stage to avoid hostile conditions, however the inducing mechanism still remains poorly understood. A similar dauer strategy exists in Caenorhabditis elegans, and dauer phenomones, which are produced through a four step cycle of peroxisomal
An Artemisinin Derivative ART1 Induces Ferroptosis by Targeting the HSD17B4 Protein Essential for Lipid Metabolism and Directly Inducing Lipid Peroxidation
Xie J, et al.
CCS Chemistry, 4(1), 304-317 (2022)
Der-Shyan Sheu et al.
Analytical biochemistry, 393(1), 62-66 (2009-06-17)
This study presents a method to detect active polyhydroxyalkanoate (PHA) synthase on a polyacrylamide gel that combines the polyhydroxybutyrate (PHB) polymerization reaction with Sudan Black B staining. After separation of the protein samples on a modified sodium dodecyl sulfate-polyacrylamide gel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service