Skip to Content
Merck
All Photos(5)

Key Documents

P5394

Sigma-Aldrich

L-α-Phosphatidylcholine

from dried egg yolk, Type X-E, ≥40% (enzymatic)

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
5209585
EC Number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

egg yolk (dried)

Quality Level

type

Type X-E

form

solid

concentration

≥40% (enzymatic)

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Hydroxyzine- and cetirizine-loaded liposomes: effect of duration of thin film hydration, freeze-thawing, and changing buffer pH on encapsulation and stability.: Discusses the encapsulation efficiency of hydroxyzine and cetirizine in L-α-Phosphatidylcholine liposomes, emphasizing the impact of preparation conditions on liposomal stability and drug release (Elzainy et al., 2005).

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey Buter et al.
Nature chemical biology, 15(9), 889-899 (2019-08-21)
Mycobacterium tuberculosis (Mtb) is the world's most deadly pathogen. Unlike less virulent mycobacteria, Mtb produces 1-tuberculosinyladenosine (1-TbAd), an unusual terpene nucleoside of unknown function. In the present study 1-TbAd has been shown to be a naturally evolved phagolysosome disruptor. 1-TbAd
Jinsi Lin et al.
Parasites & vectors, 8, 125-125 (2015-04-17)
Clonorchis sinensis (C. sinensis) inhabits in bile duct of the host. However, the mechanisms involved in why C. sinensis can survive in the bile environment containing lipids have not yet been explored. In this study, C. sinensis acetoacetyl-CoA thiolase (CsACAT)
Cristian Dima et al.
Food chemistry, 195, 39-48 (2015-11-18)
The encapsulation by spray drying method of coriander essential oil (CEO) in various materials (chitosan, alginate, chitosan/alginate, chitosan/inulin) was studied. The viscoelastic properties of the oil-in-water (O/W) emulsions and the characteristics of CEO-loaded microcapsules like morphology, moisture, wettability, solubility, flowability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service