Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
L-α-Phosphatidylcholine
Synonym(s):
1,2-Diacyl-
sn
-glycero-3-phosphocholine, 3-
sn
-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC
CAS Number:
8002-43-5
EC Number:
232-307-2
Products
Product Category
lipids (11)
glycerides (9)
metabolites (3)
media supplements (1)
Brand
Millipore (1)
Sigma-Aldrich (10)
Biological Source
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Optical Isomer
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
P3556
L-α-Phosphatidylcholine,
egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder
Expand
View Pricing
P5638
L-α-Phosphatidylcholine,
from soybean, Type II-S, 14-29% choline basis
Expand
View Pricing
P7443
L-α-Phosphatidylcholine,
from soybean, ≥99% (TLC), lyophilized powder
Expand
View Pricing
P3644
L-α-Phosphatidylcholine,
from soybean, Type IV-S, ≥30% (enzymatic)
Expand
View Pricing
61755
L-α-Phosphatidylcholine,
egg yolk, ~60% (TLC)
Expand
View Pricing
P5394
L-α-Phosphatidylcholine,
from dried egg yolk, Type X-E, ≥40% (enzymatic)
Expand
View Pricing
P3782
L-α-Phosphatidylcholine,
solution, from soybean, suitable for cell culture, Type III-S
Expand
View Pricing
P2772
L-α-Phosphatidylcholine,
egg yolk, Type XI-E, 100 mg/mL in chloroform, ≥99%, solution
Expand
View Pricing
P4279
L-α-Phosphatidylcholine,
egg yolk, Type XIII-E, 100 mg/mL in ethanol, solution
Expand
View Pricing
61771
L-α-Phosphatidylcholine,
from dried egg yolk, ≥50% (TLC)
Expand
View Pricing
44924
L-α-Phosphatidylcholine,
suitable for microbiology, Surfactant and constituent of the liposomal carrier system
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More